Skip to Content
Merck
All Photos(5)

Key Documents

HPA018519

Sigma-Aldrich

Anti-RFX5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNA-binding protein RFX5, Anti-Regulatory factor X 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ARRLGGRGQSKYCYSGIRRKTLVSMPPLPGLDLKGSESPEMGPEVTPAPRDELVEAACALTCDWAERILKRSFSSIVEVARFLLQQHLISARSAHAHVLKAMGLAEEDEHAPRERSSKPKNGLENPEGGAHKKPERLAQPPKDLEART

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RFX5(5993)

General description

The gene regulatory factor X-5 (RFX5) is mapped to human chromosome 1q21. RFX5 contains a DNA-binding domain. The protein is mainly present in the nucleus.

Immunogen

DNA-binding protein RFX5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Regulatory factor X-5 (RFX5) is a subunit of the RFX complex. The RFX complex binds X and S boxes in major histocompatibility complex class II (MHC II) promoters and regulate expression of MHC II. Mutation in RFX5 is associated with BLS (Bare lymphocyte syndrome). Transactivator CIITA (MHC class II transactivator) is important for regulation of interferon γ (IFN-γ)-induced MHC II activation and type I collagen repression. IFN-γ represses collagen transcription partly via the induction of RFX5 complex proteins and CIITA to the collagen transcription start site.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78187

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The bare lymphocyte syndrome: molecular clues to the transcriptional regulation of major histocompatibility complex class II genes.
A DeSandro et al.
American journal of human genetics, 65(2), 279-286 (1999-07-27)
N Nekrep et al.
Molecular and cellular biology, 20(12), 4455-4461 (2000-05-29)
The regulatory factor X (RFX) complex, which contains RFXANK(B), RFXAP, and RFX5, binds to X and S boxes in major histocompatibility complex class II (MHC II) promoters. In the bare lymphocyte syndrome (BLS), which is a human severe combined immunodeficiency
Nada Nekrep et al.
Nature immunology, 3(11), 1075-1081 (2002-10-09)
Bare lymphocyte syndrome (BLS) is an autosomal recessive severe-combined immunodeficiency that can result from mutations in four different transcription factors that regulate the expression of major histocompatibility complex (MHC) class II genes. We have identified here the defective gene that
Jun Xia et al.
Biochemical and biophysical research communications, 428(2), 264-270 (2012-10-20)
Decreased expression of collagen by vascular smooth muscle cells (SMCs) within the atherosclerotic plaque contributes to the thinning of the fibrous cap and poses a great threat to plaque rupture. Elucidation of the mechanism underlying repressed collagen type I (COL1A2)
Xiaocen Kong et al.
Journal of molecular and cellular cardiology, 46(3), 292-299 (2008-12-02)
Macrophage-triggered chronic inflammation and smooth muscle cell-initiated vascular remodeling are two major pathophysiologic events during atherogenesis. Major histocompatibility class II (MHC II) transactivator (CIITA) is a key mediator of these processes through transcriptional regulation of interferon gamma (IFN-gamma) induced MHC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service