Skip to Content
Merck
All Photos(4)

Documents

HPA018434

Sigma-Aldrich

Anti-CBR3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Carbonyl reductase [NADPH] 3, Anti-NADPH-dependent carbonyl reductase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DDPMPFDIKAEMTLKTNFFATRNMCNELLPIMKPHGRVVNISSLQCLRAFENCSEDLQERFHSETLTEGDLVDLMK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBR3(874)

General description

The gene CBR3 (carbonyl reductase [NADPH] 3) is mapped to human chromosome 21q22.2. CBR3 is ubiquitously expressed and is localized in the cytoplasm of HEK293 cells.

Immunogen

Carbonyl reductase [NADPH] 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Carbonyl reductase [NADPH] 3 (CBR3) catalyzes reduction of anthracyclines to cardiotoxic alcohol metabolites. Single nucleotide polymorphism in CBR3 is associated with type 2 diabetes due to impairment in prostaglandin E2 to prostaglandin F2α conversion. CBR3 expression is regulated via NRF2 (Nuclear factor erythroid 2-related factor 2). In addition, pro-inflammatory stimuli (tumor necrosis factor-α, interleukin-1β and lipopolysaccharide) induce the expression of the CBR3 gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73873

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Malátková et al.
Biochemical and biophysical research communications, 420(2), 368-373 (2012-03-20)
Until today, the physiologic role of human carbonyl reductase 3 (CBR3; SDR21C2), a member of the short-chain dehydrogenase/reductase superfamily remains obscure. Since the transcriptional regulation is closely related to the function of a protein, elucidation of the regulation of CBR3
Yi-Cheng Chang et al.
Journal of molecular medicine (Berlin, Germany), 90(7), 847-858 (2012-04-25)
Prostaglandins are potent modulators of insulin sensitivity. We systemically evaluated the association of 61 tag single-nucleotide polymorphisms (SNP) in 14 genes involved in prostaglandin metabolism with type 2 diabetes. Among all genotyped SNPs, rs10483032 in the CBR3 (carbonyl reductase 3)
Javier G Blanco et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 30(13), 1415-1421 (2011-11-30)
Carbonyl reductases (CBRs) catalyze reduction of anthracyclines to cardiotoxic alcohol metabolites. Polymorphisms in CBR1 and CBR3 influence synthesis of these metabolites. We examined whether single nucleotide polymorphisms in CBR1 (CBR1 1096G>A) and/or CBR3 (CBR3 V244M) modified the dose-dependent risk of
K Watanabe et al.
Genomics, 52(1), 95-100 (1998-09-19)
To find the genes contributing to Down syndrome, we constructed a 4-Mb sequence-ready map spanning chromosome 21q22.2 with megabase-sized cosmid/P1-derived artificial chromosome (PAC) contigs. The restriction map with rare cutting enzymes, followed by sequencing from the clustering sites, has defined
Takeshi Miura et al.
Molecular and cellular biochemistry, 315(1-2), 113-121 (2008-05-22)
Monomeric carbonyl reductases (CBRs) are enzymes that catalyze the reduction of many endogenous and xenobiotic carbonyl compounds, including steroids and prostaglandins. There are two monomeric CBR genes in the human genome, cbr1 and cbr3, which exhibit high homology in their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service