Skip to Content
Merck
All Photos(8)

Key Documents

HPA017013

Sigma-Aldrich

Anti-ZNF207 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein 207

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

RNAi knockdown
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KPATLTTTSATSKLIHPDEDISLEERRAQLPKYQRNLPRPGQAPIGNPPVGPIGGMMPPQPGIPQQQGMRPPMPPHGQYGGHHQGMPGYLPGAMPPYGQGP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF207(7756)

General description

ZNF207 (zinc finger protein 207) is a microtubule-associated zinc finger protein with molecular mass of 50.7kDa. It is expressed in several human tissues including brain, breast, CNS, colon, foreskin, germ cell, heart, liver, lung, muscle, prostate, tonsil, uterus, and embryo. Structurally, it consists of nearly 20% proline residues a potential nuclear localization signal at the amino terminus and two C2-H2 zinc finger motifs.

Immunogen

Zinc finger protein 207 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF207 (zinc finger protein 207) is involved in the equal chromosome segregation. It mainly binds to the spindle microtubules via its N-terminal domain and regulates chromosome alignment. It also acts as a potential transcription factor with DNA binding capacity. It has been reported that ZNF207 possesses ability to control transcription of β-Gal reporter gene in HeLa cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70839

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Jiang et al.
Developmental cell, 28(3), 268-281 (2014-01-28)
Equal chromosome segregation requires proper assembly of many proteins, including Bub3, onto kinetochores to promote kinetochore-microtubule interactions. By screening for mitotic regulators in the spindle envelope and matrix (Spemix), we identify a conserved Bub3 interacting and GLE-2-binding sequence (GLEBS) containing
ZNF207, a ubiquitously expressed zinc finger gene on chromosome 6p21.3.
P M Pahl et al.
Genomics, 53(3), 410-412 (1998-11-04)
Yihan Wan et al.
The Journal of cell biology, 209(2), 235-246 (2015-04-29)
Although studies suggest that perturbing mitotic progression leads to DNA damage and p53 activation, which in turn lead to either cell apoptosis or senescence, it remains unclear how mitotic defects trigger p53 activation. We show that BuGZ and Bub3, which
Yuejia Huang et al.
The Journal of cell biology, 217(1), 107-116 (2017-10-28)
Protein phase separation or coacervation has emerged as a potential mechanism to regulate biological functions. We have shown that coacervation of a mostly unstructured protein, BuGZ, promotes assembly of spindle and its matrix. BuGZ in the spindle matrix binds and
Alexander F Chin et al.
Protein science : a publication of the Protein Society, 31(4), 822-834 (2022-01-06)
Intrinsically disordered proteins (IDPs) effect biological function despite their sequence-encoded lack of preference for stable three-dimensional structure. Among their many functions, IDPs form membraneless cellular compartments through liquid-liquid phase separation (LLPS), also termed biomolecular condensation. The extent to which LLPS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service