Skip to Content
Merck
All Photos(8)

Key Documents

HPA015273

Sigma-Aldrich

Anti-RIPK2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CARD-containing IL-1 beta ICE-kinase, Anti-CARD-containing interleukin-1 beta-converting enzyme- associated kinase, Anti-RIP-2, Anti-RIP-like-interacting CLARP kinase, Anti-Receptor-interacting protein 2, Anti-Receptor-interacting serine/threonine-protein kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LPAPQDNDFLSRKAQDCYFMKLHHCPGNHSWDSTISGSQRAAFCDHKTTPCSSAIINPLSTAGNSERLQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIPK2(8767)

General description

RIPK2 (receptor-interacting serine-threonine kinase 2) is a member of the kinase family called RIP, which has seven members. It is an intracellular protein, and its C-terminal contains CARD (caspase activation and recruit domain). This gene is localized to human chromosome 8q21, and codes for a protein composed of 531 amino acids, and with a molecular weight of 61kDa. An alternatively spliced variant of this gene is called RIP2β, which lacks the C-terminal CARD and the intermediate region. It only contains a part of the kinase domain present at the N-terminal.

Immunogen

Receptor-interacting serine/threonine-protein kinase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RIPK2 (receptor-interacting serine-threonine kinase 2) plays a role in both innate and adaptive immune pathways. It also interacts with T-cell receptor (TCR) complex, and is a part of the signaling pathway. It is a part of the Toll-like receptor (TLR), Nod, IL (interleukin)-1R receptors and IL-12 transduction pathways. Studies in Chinese population suggest that variants in this gene might be linked to susceptibility to systemic lupus erythematosus (SLE). It stimulates the activity of caspase-8 and thus, promotes CD95-mediated apoptosis. It facilitates the maturation of IL-1β, by interacting with caspase-1. It is essential for the proliferation of keratinocytes. In glucocorticoid-treated patients, its expression is suppressed, which leads to decreased re-epithelialization of wounds. It plays a part in the tumorigenesis of human colon cancer, and might have potential as a marker for the aggressiveness of human colon cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73447

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Adams et al.
Experimental cell research, 316(5), 728-736 (2009-12-23)
We could recently demonstrate an important role of receptor interacting protein 4 (RIP4) in the regulation of keratinocyte differentiation. Now, we analyzed a potential role of the RIP4 homolog RIP2 in keratinocytes. Specifically, we demonstrate here that rip2 expression is
Jing Li et al.
Mutagenesis, 27(3), 319-322 (2011-11-15)
The aim of this study was to investigate the association of receptor interacting protein 2 (RIP2) single-nucleotide polymorphisms (SNPs) with susceptibility to systemic lupus erythematosus (SLE) in a Chinese population. A case-control study was performed on the SNPs rs16900617 and
Andreas Krieg et al.
Molecular immunology, 46(6), 1163-1170 (2009-01-06)
RIP2/RICK/CARDIAK is a member of the receptor interacting protein kinase (RIP) family. RIP2 promotes NF-kappaB activation as well as activation of the MAPKs JNK, ERK1/2 and p38 MAPK, thereby playing an emergent role in the innate immune response and NOD
S Groeger et al.
Infection and immunity, 88(5) (2020-02-12)
Programmed death-ligand 1 (PD-L1/B7-H1) serves as a cosignaling molecule in cell-mediated immune responses and contributes to chronicity of inflammation and the escape of tumor cells from immunosurveillance. Here, we investigated the molecular mechanisms leading to PD-L1 upregulation in human oral
Hannah Flebbe et al.
Cancers, 11(8) (2019-08-14)
Epigenetic alterations play a central role in cancer development and progression. The acetylation of histone 3 at lysine 27 (H3K27ac) specifically marks active genes. While chromatin immunoprecipitation (ChIP) followed by next-generation sequencing (ChIP-seq) analyses are commonly performed in cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service