Skip to Content
Merck
All Photos(7)

Documents

HPA014967

Sigma-Aldrich

Anti-SLC12A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2, Anti-Kidney-specific Na-K-Cl symporter, Anti-Solute carrier family 12 member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC12A1(6557)

General description

SLC12A1 (solute carrier family 12, member 1) is a member of SLC12A family of electrically neutral cation-chloride cotransporters, which contains both NaCl and KCl co-transporters. It is a bumetanide-sensitive Na-K-2Cl cotransporter and is also called NKCC2. This gene is localized to human chromosome 15 (NKCC2). This transporter resides predominantly in the plasma membrane, and has three different isoforms namely, NKCC2B, NKCC2A and NKCC2F. It has twelve transmembrane helices, with its N-terminal facing the cytoplasm. Its transport domain is made of two structurally homologous domains, made of 5-helices each. It has a large C-terminal, present intracellularly, which is responsible for dimerization of the protein.

Immunogen

Solute carrier family 12 member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC12A1 (solute carrier family 12, member 1) plays a key role in blood pressure homeostasis, and has a predominant role in the transport of salt in the thick ascending limb of Henle′s loop. It is responsible for the simultaneous movement of Na+, 1 K+, and 2 Cl- across the plasma membrane. Elevated activity of this transporter leads to inherited hypertension. Mutations in this gene are linked to type I Bartter syndrome (BS), which is characterized by hypokalaemia, excess volume depletion, and metabolic alkalosis usually leading to prenatal fatality. Mutations in this gene are also linked to atypical type I Bartter syndrome, which also includes focal segmental glomerulosclerosis. Rare mutations in this gene lead to decreased salt reabsorption in kidney, which results in hypotension.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73255

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin J Lee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(11), e21982-e21982 (2021-10-26)
Activation of the basolateral calcium sensing receptor (CaSR) in the renal tubular thick ascending limb (TAL) increases claudin-14 expression, which reduces paracellular calcium (Ca2+ ) permeability, thus increasing urinary Ca2+ excretion. However, the upstream signaling pathway contributing to altered CLDN14
Patrick G J F Starremans et al.
Journal of the American Society of Nephrology : JASN, 14(6), 1419-1426 (2003-05-23)
Bartter syndrome (BS) is a heterogeneous renal tubular disorder affecting Na-K-Cl reabsorption in the thick ascending limb of Henle's loop. BS type I patients typically present with profound hypokalemia and metabolic alkalosis. The main goal of the present study was
I Carota et al.
Acta physiologica (Oxford, England), 199(3), 327-338 (2010-02-12)
Salt reabsorption across the apical membrane of cells in the thick ascending limb (TAL) of Henle is primarily mediated by the bumetanide-sensitive Na(+)/K(+)/2Cl(-) cotransporter NKCC2. Three full-length splice variants of NKCC2 (NKCC2B, NKCC2A and NKCC2F) have been described. The NKCC2
Hajime Yamazaki et al.
Pediatric nephrology (Berlin, Germany), 24(2), 415-418 (2008-10-03)
Type I Bartter syndrome (BS) is caused by mutations of the Na-K-2Cl cotransporter (NKCC2)-encoding SLC12A1 gene. The clinical phenotype of this severe form of BS is characterized by polyhydramnios, premature delivery, failure to thrive, and nephrocalcinosis, and the diagnosis is
Michelle Y Monette et al.
American journal of physiology. Renal physiology, 300(4), F840-F847 (2011-01-07)
The Na-K-Cl cotransporter (NKCC2) is the major salt transport pathway in the thick ascending limb of Henle's loop and is part of the molecular mechanism for blood pressure regulation. Recent screening of ∼3,000 members of the Framingham Heart Study identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service