Skip to Content
Merck
All Photos(4)

Key Documents

HPA014788

Sigma-Aldrich

Anti-BVES antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Blood vessel epicardial substance, Anti-Popeye domain-containing protein 1, Anti-Popeye protein 1, Anti-hBVES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LNDPTLNDKKAKKLEHQLSLCTQISMLEMRNSIASSSDSDDGLHQFLRGTSSMSSLHVSSPHQRASAKMKPIEEGAEDDDDVFEPASPNTLKVHQLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BVES(11149)

General description

BVES (blood vessel epicardial substance) is the prototypic member of Popeye Domain Containing (Popdc) family, and is also called POPDC1. This family has two other members called POPDC2 and POPDC3. BVES is a transmembrane epithelial adhesion protein, containing the characteristic Popeye domain made of amino acids 172-266. It spans the membrane 3 times, and its C-terminal faces the cytoplasm. Homotypic interaction of BVES-BVES is essential for its localization to plasma membrane. It was initially identified in the cDNA library of developing heart.

Immunogen

Blood vessel epicardial substance recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BVES (blood vessel epicardial substance) is involved in the control of tight junction formation in epithelial cells. These tight junctions in turn control RhoA and ZONAB/DbpA, which is a y-box transcription factor. Therefore, the expression and residence of BVES has a regulatory effect on RhoA and ZONAB/DbpA activity. Suppression of this protein leads to DNA hypermethylation or the silencing of transcription. It is down-regulated in all stages of human colorectal carcinoma (CRC) and in adenomatous polyps. Its down-regulation is associated with epithelial-mesenchymal transition (EMT), and consequent colon tumorigenesis. BVES is also down-regulated in gastric cancer, and this is associated with enhanced tumorigenesis and poor patient prognosis. This protein is a key player in the development of embryo and cardiocyte differentiation. It is over-expressed in patients with congenital defects of septa and is linked with tetralogy of Fallot (TOF).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73021

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willem De Ridder et al.
Neurology. Genetics, 5(2), e321-e321 (2019-05-24)
To study the genetic and phenotypic spectrum of patients harboring recessive mutations in BVES. We performed whole-exome sequencing in a multicenter cohort of 1929 patients with a suspected hereditary myopathy, showing unexplained limb-girdle muscular weakness and/or elevated creatine kinase levels.
Min Wu et al.
International journal of molecular medicine, 31(4), 899-903 (2013-02-14)
Tetralogy of Fallot (TOF) is a common congenital heart defect (CHD). However, the genetic causes are largely unknown. Blood vessel epicardial substance (BVES) is postulated to play a role in embryonic development, and we previously found that the expression of
Tanya A Baldwin et al.
EMBO reports, 23(12), e55208-e55208 (2022-10-19)
The establishment of macromolecular complexes by scaffolding proteins is key to the local production of cAMP by anchored adenylyl cyclase (AC) and the subsequent cAMP signaling necessary for cardiac functions. We identify a novel AC scaffold, the Popeye domain-containing (POPDC)
Deng Luo et al.
Pathology oncology research : POR, 18(2), 491-497 (2011-11-24)
Although many molecular and biological studies have shown risk factors for gastric cancer, the available knowledge is still insufficient to unveil the exact mechanism of gastric cancer. To investigate the relationships between Bves expression and the clinicopathologic features of gastric
Haiwen Li et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 31(2), 398-408 (2022-11-27)
Limb-girdle muscular dystrophy type R25 (LGMDR25) is caused by recessive mutations in BVES encoding a cAMP-binding protein, characterized by progressive muscular dystrophy with deteriorating muscle function and impaired cardiac conduction in patients. There is currently no therapeutic treatment for LGMDR25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service