Skip to Content
Merck
All Photos(8)

Key Documents

HPA013144

Sigma-Aldrich

Anti-VAPB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-VAMP-B/VAMP-C, Anti-VAMP-associated protein B/C, Anti-VAP-B/VAP-C, Anti-Vesicle-associated membrane protein-associated protein B/C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VWKEAKPEDLMDSKLRCVFELPAENDKPHDVEINKIISTTASKTETPIVSKSLSSSLDDTEVKKVMEECKRLQGEVQRLREENKQFKEEDGLRMRKTVQSNSPISALAPTGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAPB(9217)

General description

Vesicle-associated membrane protein-associated protein B/C (VAPB) belongs to the VAP family. It contains three structural domains: the major sperm protein (MSP) domain, an amphipathic helical structure and a carboxy-terminal domain that acts as a membrane anchor. VAPB is localized in intracellular membranes of the endoplasmic reticulum and Golgi apparatus. The gene encoding this protein is located on chromosome 20q13.3.

Immunogen

Vesicle-associated membrane protein-associated protein B/C recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Vesicle-associated membrane protein-associated protein B/C (VAPB) plays a role during endoplasmic reticulum (ER)-Golgi transport and secretion. It has been shown to act in the unfolded protein response which is activated whenever misfolded proteins accumulate in the ER. Mutations in the gene encoding VAPB are associated with amyotrophic lateral sclerosis and late-onset spinal muscular atrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71876

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu Kui Gao et al.
Cell discovery, 9(1), 83-83 (2023-08-02)
The scaffold protein IRS-1 is an essential node in insulin/IGF signaling. It has long been recognized that the stability of IRS-1 is dependent on its endomembrane targeting. However, how IRS-1 targets the intracellular membrane, and what type of intracellular membrane
Han-Jou Chen et al.
The Journal of biological chemistry, 285(51), 40266-40281 (2010-10-14)
Following the mutation screening of genes known to cause amyotrophic lateral sclerosis (ALS) in index cases from 107 familial ALS (FALS) kindred, a point mutation was identified in vesicle-associated membrane protein-associated protein B (VAPB), or VAMP-associated protein B, causing an
Fumiaki Mori et al.
Brain pathology (Zurich, Switzerland), 31(6), e13001-e13001 (2021-07-02)
The pathological hallmark of multiple system atrophy (MSA) is fibrillary aggregates of α-synuclein (α-Syn) in the cytoplasm and nucleus of both oligodendrocytes and neurons. In neurons, α-Syn localizes to the cytosolic and membrane compartments, including the synaptic vesicles, mitochondria, and
Suzan Kors et al.
Methods in molecular biology (Clifton, N.J.), 2643, 345-357 (2023-03-24)
Organelles physically interact with each other via protein tethering complexes that bridge the opposing membranes. Organelle membrane contacts are highly dynamic, implying dynamism of the tethering complexes. Alterations in the binding of the tethering proteins can be assessed by immunoprecipitation.
Golam T Saffi et al.
PloS one, 16(11), e0259313-e0259313 (2021-11-24)
Lysosomes are terminal, degradative organelles of the endosomal pathway that undergo repeated fusion-fission cycles with themselves, endosomes, phagosomes, and autophagosomes. Lysosome number and size depends on balanced fusion and fission rates. Thus, conditions that favour fusion over fission can reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service