Skip to Content
Merck
All Photos(4)

Documents

HPA013138

Sigma-Aldrich

Anti-PENK antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Proenkephalin A precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

ECSQDCATCSYRLVRPADINFLACVMECEGKLPSLKIWETCKELLQLSKPELPQDGTSTLRENSKPEESHLLAKRYGGFMKRYGGFMKKMDELYPMEPEEEANGSEILAKRYGGFMKKDAEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PENK(5179)

General description

Proenkephalin-A precursor (PENK) is the prohormone precursor. The gene encoding it is localized on human chromosome 8q23-q24.

Immunogen

Proenkephalin A precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PENK antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Opioid peptides are derived from the genes encoding precursors like proenkephalin-A (PENK). It is processed at 12 cleavage sites to give rise to enkephalin peptides. These peptides take part in controlling learning and memory, analgesia and other brain-regulated behaviors. The gene is shown to be hypermethylated in prostate cancer and thus may be of use as a molecular marker for prostate cancer diagnosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71601

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia Ashour et al.
The Prostate, 74(12), 1171-1182 (2014-06-26)
DNA hypermethylation has emerged as a novel molecular biomarker for the evaluation of prostate cancer diagnosis and prognosis. Defining the specific gene hypermethylation profile for prostate cancer could involve groups of genes that specifically discriminate patients with indolent and aggressive
Guangwei Wang et al.
Journal of ovarian research, 17(1), 58-58 (2024-03-14)
Tumors are highly heterogeneous, and within their parenchyma, a small population of tumor-stem cells possessing differentiation potential, high oncogenicity, and self-renewal capabilities exists. These cells are pivotal in mediating tumor development, chemotherapy resistance, and recurrence. Ovarian cancer shares characteristics with
Weiya D Lu et al.
Protein science : a publication of the Protein Society, 21(2), 178-187 (2011-11-22)
Active enkephalin and related peptide hormones or neurotransmitters are generated by proteolytic processing of inactive prohormone precursors. Little is known about the relative accessibilities of prohormone cleavage sites and conformations of subdomains that undergo proteolytic processing. Therefore, this study investigated
M Litt et al.
American journal of human genetics, 42(2), 327-334 (1988-02-01)
DNA probes derived from rat and human proenkephalin and prodynorphin genes have been used to localize these two opiate neuropeptide genes on human chromosomes. Hybridization of probes to Southern blots made with DNAs from a rodent-human somatic-cell hybrid panel indicates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service