Skip to Content
Merck
All Photos(8)

Key Documents

HPA012924

Sigma-Aldrich

Anti-ARHGEF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-115 kDa guanine nucleotide exchange factor, Anti-Rho guanine nucleotide exchange factor 1, Anti-Sub15, Anti-p115-RhoGEF, Anti-p115RhoGEF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

YMRHLGVRTKSGDKKSGRNFFRKKVMGNRRSDEPAKTKKGLSSILDAARWNRGEPQVPDFRHLKAEVDAEKPGATDRKGGVGMPSRDRNIGAPGQDTPGVSLHPLSLDSPDREP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF1(9138)

General description

Rho guanine nucleotide exchange factor 1 (ARHGEF1) is a member of a family of Rho-specific guanine nucleotide exchange factors. It contains a regulator of G protein signaling (rgRGS) domain near its amino-terminus. ARHGEF1 also has a tandem Dbl homology (DH)-Pleckstrin homology (PH) domain.

Immunogen

Rho guanine nucleotide exchange factor 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Rho guanine nucleotide exchange factor 1(ARHGEF1) mediates dissociation of GDP and the binding of GTP. It binds specifically to the nucleotide-depleted state of the substrate. ARHGEF1 acts as a regulator of Rho and plays an important role in the activation of Ras-like proteins. Its amino terminal region helps it to function as an intermediate in the regulation of Rho proteins by G protiens G13 and G12. The GTPase activities of G12 and G13 are stimulated by it. Once G13 is activated by ARHGEF1, it catalyzes the nucleotide exchange on Rho. In arterial smooth muscle cells, it is responsible for angiotensin II-mediated activation of RhoA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72470

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of p115RhoGEF Ser(330) regulates its RhoGEF activity.
CR Chow
Cellular Signalling, 25(11), 2085-2092 (2013)
M J Hart et al.
Science (New York, N.Y.), 280(5372), 2112-2114 (1998-06-26)
Signaling pathways that link extracellular factors to activation of the monomeric guanosine triphosphatase (GTPase) Rho control cytoskeletal rearrangements and cell growth. Heterotrimeric guanine nucleotide-binding proteins (G proteins) participate in several of these pathways, although their mechanisms are unclear. The GTPase
Z Chen et al.
Nature structural biology, 8(9), 805-809 (2001-08-29)
p115RhoGEF, a guanine nucleotide exchange factor for Rho GTPase, is also a GTPase activating protein (GAP) for G(12) and G(13) heterotrimeric G alpha subunits. Near its N-terminus, p115RhoGEF contains a domain (rgRGS) with remote sequence identity to RGS (regulators of
Christophe Guilluy et al.
Nature medicine, 16(2), 183-190 (2010-01-26)
Hypertension is one of the most frequent pathologies in the industrialized world. Although recognized to be dependent on a combination of genetic and environmental factors, its molecular basis remains elusive. Increased activity of the monomeric G protein RhoA in arteries
M J Hart et al.
The Journal of biological chemistry, 271(41), 25452-25458 (1996-10-11)
The Rho GTPase promotes proliferation and cytoskeletal rearrangements in mammalian cells. To understand the regulation of Rho, it is important to characterize guanine nucleotide exchange factors (GEFs), which stimulate the dissociation of GDP and subsequent binding of GTP. Using Rho

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service