Skip to Content
Merck
All Photos(3)

Key Documents

HPA009081

Sigma-Aldrich

Anti-CDHR5 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-μ-Protocadherin, Anti-MUCDHL, Anti-MUPCDH, Anti-Mucin and cadherin-like protein precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

CIQAQYHGAVPTGHILPSPLVLRPGPIYAEDGDRGINQPIIYSIFRGNVNGTFIIHPDSGNLTVARSVPSPMTFLLLVKGQQADLARYSVTQVTVEAVAAAGSPPRFPQSLYRGTVARGAGAGVVVKDA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUPCDH(53841)

Looking for similar products? Visit Product Comparison Guide

General description

CDHR5 (cadherin related family member 5) is a protocadherin (PCDH) family member, and is also known as PCDH24. It might be a constituent of enterocyte microvillus. It is a poorly characterized protein. It is a type I transmembrane protein, which contains the characteristic domains of the cadherin protein family- extracellular EC repeats. This gene localizes to human chromosome 11p15.5.

Immunogen

Mucin and cadherin-like protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CDHR5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CDHR5 (cadherin related family member 5) is localized to the brush border, and might be involved in the adhesion of microvilli, eventually resulting in the tight packing of fully differentiated enterocytes. It interacts in a trans-fashion with MLPCDH (mucin-like protocadherin) to form Ca2+-dependent adhesion complexes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71708

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott W Crawley et al.
Cell, 157(2), 433-446 (2014-04-15)
Transporting epithelial cells build apical microvilli to increase membrane surface area and enhance absorptive capacity. The intestinal brush border provides an elaborate example with tightly packed microvilli that function in nutrient absorption and host defense. Although the brush border is
Leslie M Meenderink et al.
Developmental cell, 50(5), 545-556 (2019-08-06)
Transporting epithelial cells generate arrays of microvilli, known as a brush border, to enhance functional capacity. To understand brush border formation, we used live cell imaging to visualize apical remodeling early in this process. Strikingly, we found that individual microvilli
Michael Goldberg et al.
American journal of physiology. Renal physiology, 283(3), F454-F463 (2002-08-09)
We recently cloned mu-protocadherin, a developmentally regulated cell adhesion molecule that contains an extracellular region with four cadherin-like ectodomains and a triply repeating mucin domain in its longer isoform. Expression of mu-protocadherin in L929 cells resulted in cellular aggregation, confirming
Meredith L Weck et al.
The Journal of biological chemistry, 295(48), 16191-16206 (2020-10-15)
Solute transporting epithelial cells build arrays of microvilli on their apical surface to increase membrane scaffolding capacity and enhance function potential. In epithelial tissues such as the kidney and gut, microvilli are length-matched and assembled into tightly packed "brush borders,"
Russell E McConnell et al.
American journal of physiology. Gastrointestinal and liver physiology, 300(5), G914-G926 (2011-02-19)
The brush border domain at the apex of intestinal epithelial cells is the primary site of nutrient absorption in the intestinal tract and the primary surface of interaction with microbes that reside in the lumen. Because the brush border is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service