Skip to Content
Merck
All Photos(2)

Key Documents

HPA008723

Sigma-Aldrich

Anti-FOXJ2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fork head homologous X antibody produced in rabbit, Anti-Forkhead box protein J2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ELMESLRQAEQKNWTLDQHHIANLCDSLNHFLTQTGHVPPQGGTHRPPAPARIADSCALTSGKQESAMSQVNSYGHPQAPHLYPGPSPMYPIPTQDSAGYNRPAHHMVPRPSVPPPGANEEIPDDFDWDLI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXJ2(55810)

General description

FOXJ2 (forkhead box J2) is a mammalian fork head transcription factor, which has a wide range of adult tissue expression. It is also expressed during the cleavage stages of pre-implantation development. This protein is composed of 574 amino acids. This gene is localized to human chromosome 12p13.31. Its N- and C-termini contain two tyrosine residues and a region of basic amino acids, which target it to the nucleus, where it is localized. It is alternatively spliced into two variants-FHX.S and FHX.L, with FHK.L being the larger form and the more potent transactivator.

Immunogen

Forkhead box protein J2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FOXJ2 (forkhead box J2) facilitates the differentiation of endothelial progenitor cells (EPCs), during sheer stress, when acted upon by microRNA-34a (miR-34a). In breast cancer tissues, it controls the EMT (epithelial mesenchymal transition) by controlling E-caherin and vimentin. Thus, it prevents metastasis in breast cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70848

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maurizio Zuccotti et al.
Human reproduction (Oxford, England), 24(9), 2225-2237 (2009-05-30)
Our knowledge of what determines the mammalian oocyte developmental competence is meagre. By comparing the transcriptional profiles of developmentally competent surrounded nucleolus (SN) and incompetent not surrounded nucleolus (NSN) mouse MII oocytes, we recently demonstrated that Oct-4 and Stella are
María Ana Gómez-Ferrería et al.
Journal of molecular biology, 329(4), 631-644 (2003-06-06)
FOXJ2 is a fork head transcriptional activator, the expression of which starts very early in embryonic development and it is distributed widely in the adult. Here, we describe the characterization of domains that are important for its function. FOXJ2 is
C Pérez-Sánchez et al.
Journal of molecular biology, 301(4), 795-806 (2000-09-01)
Many biological phenomena are dependent on mechanisms that fine-tune the expression levels of particular genes. This can be achieved by altering the relative activity of a single transcription factor, by post-translational modifications or by interaction with regulatory molecules. An alternative
Yingying Wang et al.
Journal of cellular biochemistry, 113(8), 2729-2737 (2012-03-24)
The prognosis of breast cancer patients with metastases is generally poor, so it is essential to elucidate related molecules mechanisms. Forkhead Box J2 (FOXJ2) is a member of Forkhead Box transcription factors, many of which have been reported to participate
C Pérez-Sánchez et al.
The Journal of biological chemistry, 275(17), 12909-12916 (2000-04-25)
The HNF3/fork head family includes a large number of transcription factors that share a structurally related DNA binding domain. Fork head factors have been shown to play important roles both during development and in the adult. We now describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service