Skip to Content
Merck
All Photos(6)

Key Documents

HPA008436

Sigma-Aldrich

Anti-HJURP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-14-3-3-associated AKT substrate, Anti-Fetal liver-expressing gene 1 protein, Anti-Holliday junction recognition protein, Anti-Up-regulated in lung cancer 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
12
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TPVVQMATLTYETPQGLRIWGGRLIKERNEGEIQDSSMKPADRTDGSVQAAAWGPELPSHRTVLGADSKSGEVDATSDQEESVAWALAPAVPQSPLKNELRRKYLTQVDILLQGAEYFECAGNRAGRDVRV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HJURP(55355)

General description

Holliday junction recognition protein (HJURP) is a centromere protein A (CENP-A) histone chaperone and a chromatin assembly factor. It directly binds to the Holliday junction.

Immunogen

Holliday junction recognition protein recombinant protein epitope signature tag (PrEST)

Application

Anti-HJURP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry at a dilution of 1:500 against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting at a dilution of 1:500. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Holliday junction recognition protein (HJURP) is involved in the recruitment of centromere protein A (CENP-A) into nucleosomes at replicated centromeres. It is recruited in the form of a prenucleosomal complex during the early G1 phase of the cell cycle. The centromere targeting domain (CATD) of CENP-A is needed for the recognition of the protein. Thus it is a cell-cycle regulated protein which plays an important role in the centromeric chromatin assembly.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71334

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HJURP is a CENP-A chromatin assembly factor sufficient to form a functional de novo kinetochore.
MC Barnhart
The Journal of Cell Biology, 194(2), 229-243 (2011)
Wenfeng Huang et al.
PloS one, 11(2), e0148618-e0148618 (2016-02-11)
HJURP (Holliday Junction-Recognizing Protein) plays dual roles in DNA repair and in accurate chromosome segregation during mitosis. We examined whether the single nucleotide polymorphisms (SNPs) of HJURP were associated with the risk of occurrence of hepatocellular carcinoma (HCC) among chronic
Bayarbat Tsevegjav et al.
International journal of oncology, 60(3) (2022-02-02)
Since oral cancer (OC) is highly malignant and the efficacy of standard treatments is limited, the development of new therapeutics is urgently awaited. To identify potential molecular targets for new OC diagnosis and therapies, we screened oncoantigens by gene expression
Centromere-specific assembly of CENP-a nucleosomes is mediated by HJURP.
DR Foltz
Cell, 137(3), 472-484 (2009)
Marie de Tayrac et al.
PloS one, 8(9), e73332-e73332 (2013-09-17)
Recent studies have highlighted the heterogeneity of gliomas and demonstrated that molecular and genetic analysis could help in their classification and in the design of treatment protocols. In a previous study we have identified a 4-gene signature highly correlated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service