Skip to Content
Merck
All Photos(8)

Key Documents

HPA008209

Sigma-Aldrich

Anti-STXBP1 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-N-Sec1, Anti-Syntaxin-binding protein 1, Anti-Unc-18 homolog, Anti-Unc-18-1, Anti-Unc-18A, Anti-p67

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TDSCPDALFNELVKSRAAKVIKTLTEINIAFLPYESQVYSLDSADSFQSFYSPHKAQMKNPILERLAEQIATLCATLKEYPAVRYRGEYKDNALLAQLIQDKLDAYKADDPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STXBP1(6812)

General description

Syntaxin-binding protein 1 (STXBP1) belongs to the Sec1/Munc-18 (SM) family of proteins. It is an intracellular protein present in presynaptic terminals.

Immunogen

Syntaxin-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Syntaxin-binding protein 1 (STXBP1) is required for secretion of neurotransmitters and for vesicle fusion in eukaryotic cells. In neurons, Mint 1 and 2 proteins bind to STXBP1 to form the essential synaptic vesicle fusion machinery and help in regulated exocytosis. It also forms a tight complex with the Soluble NSF attachment protein receptor (SNARE) syntaxin 1, in which syntaxin is held in a closed conformation and further stabilized to form the SNARE complexes. This shows that STXBP1 allows the formation of a complex between syntaxin and Synaptosomal-Associated Protein 25kDa (SNAP-25), which in turn functions as an acceptor for vesicle-bound synaptobrevin. All these activities are essential intermediates in the pathway towards exocytosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71578

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Yang et al.
Neuron, 28(2), 375-383 (2001-01-06)
Rasmussen's encephalitis (RE) is a rare disease of the central nervous system characterized by severe epileptic seizures, progressive degeneration of a single cerebral hemisphere, and autoimmunity directed against glutamate receptor subunit, GluR3. We report here the identification of high-titer autoantibodies
John Togneri et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(47), 17730-17735 (2006-11-09)
The Sec1/Munc-18 (SM) family of proteins is required for vesicle fusion in eukaryotic cells and has been linked to the membrane-fusion proteins known as soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs). SM proteins may activate the target-membrane SNARE, syntaxin, for
Felipe E Zilly et al.
PLoS biology, 4(10), e330-e330 (2006-09-28)
Munc18-1, a protein essential for regulated exocytosis in neurons and neuroendocrine cells, belongs to the family of Sec1/Munc18-like (SM) proteins. In vitro, Munc18-1 forms a tight complex with the SNARE syntaxin 1, in which syntaxin is stabilized in a closed
M Okamoto et al.
European journal of cell biology, 77(3), 161-165 (1998-12-22)
Mint 1 and 2 are proteins that bind to munc18-1, an essential component of the synaptic vesicle fusion machinery, and are detectably expressed only in neurons [Okamoto and Südhof, J. Biol. Chem. 272, 31459-31464 (1997)]. Mint 1 and 2 are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service