Skip to Content
Merck
All Photos(15)

Key Documents

HPA007305

Sigma-Aldrich

Anti-CALB2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-29 kDa calbindin antibody produced in rabbit, Anti-CR antibody produced in rabbit, Anti-Calretinin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
independent
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:5000-1:10000

immunogen sequence

LKGFLSDLLKKANRPYDEPKLQEYTQTILRMFDLNGDGKLGLSEMSRLLPVQENFLLKFQGMKLTSEEFNAIFTFYDKDRSGYIDEHELDALLKDLYEKNKKEMNIQQLTNYRKSVMSLAEAGKLYRKDLEIV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALB2(794)

Looking for similar products? Visit Product Comparison Guide

General description

CALB2 (calbindin 2) gene encodes a calcium-binding protein belonging to the troponin C superfamily. It is also called as calretinin. Structurally, members of this family consists of six EF-hand domains that bind to calcium. It has a molar mass of 29kDa. It is predominantly expressed by certain cells in the nervous system and also in ovarian cells. It is abundantly expressed in colon carcinomas. The gene is localized to human chromosome 16q22.2.

Immunogen

Calretinin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CALB2 (calbindin 2) gene encodes a protein that functions in modulating neuronal excitability and also induces apoptosis through the intrinsic mitochondrial pathway. It is involved in several cellular processes, including message targeting and intracellular calcium buffering. It serves as a diagnostic marker for Hirschsprung disease and some cancers. It also serves as a marker for mesothelial differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70178

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross J Taliano et al.
Human pathology, 44(12), 2743-2750 (2013-11-23)
Calretinin, a calcium-binding protein, is a widely used marker for mesothelial differentiation. There is accumulating evidence of calretinin expression in epithelial and mesenchymal malignancies, as well. The objectives of this study were to (1) further delineate the expression of calretinin
Jill C Wooff et al.
The American journal of surgical pathology, 35(12), 1786-1793 (2011-10-25)
Olfactory neuroblastoma (ONB) is an uncommon malignant tumor of the sinonasal tract and has a wide histologic differential diagnosis that includes other small round blue cell tumors (SRBCTs). Even with the use of immunohistochemistry (IHC), the correct diagnosis may be
Leanne Stevenson et al.
PloS one, 6(5), e20276-e20276 (2011-06-02)
The role of the calcium binding protein, Calbindin 2 (CALB2), in regulating the response of colorectal cancer (CRC) cells to 5-Fluorouracil (5-FU) was investigated. Real-time RT-PCR and Western blot analysis revealed that CALB2 mRNA and protein expression were down-regulated in
Beatrice Malacrida et al.
iScience, 24(6), 102676-102676 (2021-07-01)
Guided by a multi-level "deconstruction" of omental metastases, we developed a tetra (four cell)-culture model of primary human mesothelial cells, fibroblasts, adipocytes, and high-grade serous ovarian cancer (HGSOC) cell lines. This multi-cellular model replicated key elements of human metastases and
M Parmentier et al.
Cytogenetics and cell genetics, 57(1), 41-43 (1991-01-01)
The genes encoding calbindin D28k (CALB1) and calretinin (CALB2), two closely related calcium-binding proteins, were mapped by in situ hybridization to the 8q21.3----q22.1 and 16q22----q23 regions of the human genome, respectively. These localizations match the chromosomal regions where the carbonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service