Skip to Content
Merck
All Photos(6)

Key Documents

HPA006880

Sigma-Aldrich

Anti-SCG3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Secretogranin III antibody produced in rabbit, Anti-Secretogranin-3 precursor antibody produced in rabbit, Anti-SgIII antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LDGTPLTAEDIVHKIAARIYEENDRAVFDKIVSKLLNLGLITESQAHTLEDEVAEVLQKLISKEANNYEEDPNKPTSWTENQAGKIPEKVTPMAAIQDGLAKGE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCG3(29106)

Immunogen

Secretogranin-3 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SCG3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SCG3 (Secretogranin III) is a secretory protein belonging to the multifunctional granin family. It is widely distributed in many tissues specifically in the brain. It is transmitted from the cell through trans-Golgi network (TGN). SCG3 is majorly involved in two functions; firstly, the storage and transportation of neurotransmitters, and secondly, biogenesis of secretory granule. It also generates peptide hormones through the regulated secretory pathway. It has been reported that SCG3 may participate in the apoptotic signaling pathway. It has also been postulated that SCG3 may play a role in the neuroendocrine pathway which proves pathological implications in the Parkinson′s disease (PD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70128

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph S Canzano et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(1), 41-52 (2018-05-18)
Islet microvasculature provides key architectural and functional roles, yet the morphological features of islets from patients with type 1 diabetes are poorly defined. We examined islet and exocrine microvasculature networks by multiplex immunofluorescence imaging of pancreases from organ donors with
Sonia Paco et al.
Cerebral cortex (New York, N.Y. : 1991), 20(6), 1386-1397 (2009-11-07)
Astrocytes release peptide and nonpeptide transmitters that influence neuronal development, function, and plasticity. However, the molecular components of the astroglial secretory pathways in vivo are largely unknown. Here, we analyze in astrocytes the production, expression regulation, trafficking, and release of
Fengrui Li et al.
Journal of neuroscience research, 90(12), 2237-2246 (2012-09-19)
The neurotoxins paraquat (PQ) and dopamine (DA or 6-OHDA) cause apoptosis of dopaminergic neurons in the substantia nigra pars compacta (SNpc), reproducing an important pathological feature of Parkinson's disease (PD). Secretogranin III (SCG3), a member of the multifunctional granin family
Yi-Ping Rong et al.
Sheng wu hua xue yu sheng wu wu li xue bao Acta biochimica et biophysica Sinica, 34(4), 411-417 (2002-07-06)
We have cloned a new member of the granin family, termed human secretogranin III (SgIII), that encodes 468 amino acid residues. The human SgIII protein possesses an N-terminal signal peptide, seven dibasic sites, and the repeated DSTK sequences. These structure
Martha Campbell-Thompson et al.
Frontiers in endocrinology, 12, 644826-644826 (2021-05-14)
Although first described over a hundred years ago, tissue optical clearing is undergoing renewed interest due to numerous advances in optical clearing methods, microscopy systems, and three-dimensional (3-D) image analysis programs. These advances are advantageous for intact mouse tissues or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service