Skip to Content
Merck
All Photos(6)

Key Documents

HPA005830

Sigma-Aldrich

Anti-L1CAM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD171 antigen antibody produced in rabbit, Anti-N-CAM L1 antibody produced in rabbit, Anti-Neural cell adhesion molecule L1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

ELRTHNLTDLSPHLRYRFQLQATTKEGPGEAIVREGGTMALSGISDFGNISATAGENYSVVSWVPKEGQCNFRFHILFKALGEEKGGASLSPQYVSYNQSSYTQWDLQPDTDYEIHLFKERMFRHQMAVKTNGTGRVRLPPAGFATE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... L1CAM(3897)

General description

The gene L1CAM (L1 cell adhesion molecule) encodes a member of the immunoglobulin supergene family. The encoded protein contains a cytoplasmic intracellular domain, one single-pass transmembrane region and an extracellular domain containing six immunoglobulin- (Ig) and five fibronectin (Fn) III-like motifs. The gene is mapped to human chromosome Xq28. It contains 29 exons that encode a protein of 1257 amino acids containing a 19 amino acid signal peptide and a functional L1 protein of 1238 amino acids. It is mainly expressed in the nervous system.

Immunogen

Neural cell adhesion molecule L1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-L1CAM antibody has been used to print on epoxy slides prior to EV capturing and analysis. All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene L1CAM (L1 cell adhesion molecule) encodes a glycoprotein that functions in axon growth during development and axon bundling. It interacts with Schwann cells and axons, and mediates neuronal cell migration, and neuronal cell survival. Overexpression of this protein is associated with tumor progression. It has been found to mediate therapeutic resistance in ovarian and pancreatic cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74285

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donatella Delle Cave et al.
Theranostics, 11(12), 5686-5699 (2021-04-27)
Background: Colorectal cancer (CRC) is currently the third leading cause for cancer-related mortality. Cancer stem cells have been implicated in colorectal tumor growth, but their specific role in tumor biology, including metastasis, is still uncertain. Methods: Increased expression of L1CAM
Malene Møller Jørgensen et al.
Journal of extracellular vesicles, 4, 26048-26048 (2015-04-12)
Extracellular vesicles (EVs) and exosomes are difficult to enrich or purify from biofluids, hence quantification and phenotyping of these are tedious and inaccurate. The multiplexed, highly sensitive and high-throughput platform of the EV Array presented by Jørgensen et al., (J
Alexandra Brahmer et al.
Journal of extracellular vesicles, 8(1), 1615820-1615820 (2019-06-14)
Physical activity initiates a wide range of multi-systemic adaptations that promote mental and physical health. Recent work demonstrated that exercise triggers the release of extracellular vesicles (EVs) into the circulation, possibly contributing to exercise-associated adaptive systemic signalling. Circulating EVs comprise
L1CAM malfunction in the nervous system and human carcinomas.
Schafer MK and Altevogt P
Cellular and Molecular Life Sciences, 67, 2425-2437 (2010)
An updated and upgraded L1CAM mutation database.
Vos YJ and Hofstra RM
Human Mutation, 31, E1102-E1109 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service