Skip to Content
Merck
All Photos(2)

Key Documents

HPA005661

Sigma-Aldrich

Anti-ADAMTS5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A disintegrin and metalloproteinase with thrombospondin motifs 5 antibody produced in rabbit, Anti-ADAM-TS 11 antibody produced in rabbit, Anti-ADAM-TS 5 antibody produced in rabbit, Anti-ADAM-TS5 antibody produced in rabbit, Anti-ADAMTS-5 precursor antibody produced in rabbit, Anti-ADMP-2 antibody produced in rabbit, Anti-Aggrecanase-2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KGLVQNIDQLYSGGGKVGYLVYAGGRRFLLDLERDGSVGIAGFVPAGGGTSAPWRHRSHCFYRGTVDGSPRSLAVFDLCGGLDGFFAVKHARYTLKPLLRGPWAEEEKGRVYGDGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAMTS5(11096)

General description

ADAM metallopeptidase with thrombospondin motifs 5 (ADAMTS5) belongs to the ADAM protein family. It has an ADAM-like protease domain, a disintegrin-like domain and a cysteine-rich domain. ADAMTS5 lacks a transmembrane domain and is thus secreted into the extracellular matrix.

Immunogen

A disintegrin and metalloproteinase with thrombospondin motifs 5 precursor (ADAMTS-5) (ADAM-TS5) (Aggrecanase-2) (ADMP-2) (A disintegrin and metalloproteinase with thrombospondin motifs 11) (ADAMTS-11)

Application

Anti-ADAMTS5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ADAM metallopeptidase with thrombospondin motifs 5 (ADAMTS5) is involved in aggrecan, brevican and α2-macroglobulin degradation. It is one of the most important extra cellular matrix (ECM) degrading enzymes and functions in tissue degradation, remodeling and cell infiltration. ADAMTS5 cleaves cartilage aggrecan at the Glu (373)-Ala (374) bond and also in the region spanning residues Gly (1481) and Glu (1667), which represents its unique cleavage site.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85176

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyuan Gong et al.
Journal of cellular physiology, 234(6), 9711-9722 (2018-10-30)
Ca2+ has been recognized as a key molecule for chondrocytes, however, the role and mechanism of spontaneous [Ca 2+ ] i signaling in cartilaginous extracellular matrix (ECM) metabolism regulation are unclear. Here we found that spontaneous Ca 2+ signal of
Expression of ADAMTS-5/implantin in human decidual stromal cells: regulatory effects of cytokines.
H. Zhu
Human Reproduction, 22(1), 63-74 (2007)
Priscilla B Pail et al.
International immunopharmacology, 72, 62-73 (2019-04-09)
This study evaluated the role of kinin B1 and B2 receptors in the pre-clinical mouse model of oxazolone-induced atopic dermatitis. The B1 R715 or B2 HOE140 receptor antagonists were dosed at different schemes of treatment. After assessment of clinical lesion
Tao Wang et al.
International journal of molecular medicine, 44(2), 630-642 (2019-06-15)
Osteoarthritis (OA) is a common and troublesome disease among the elderly, and is characterized by extracellular matrix (ECM) degradation. The function of the long non‑coding RNA X‑inactive‑specific transcript (XIST) and its working mechanism in ECM degradation remains unclear. In the
Cheng Yang et al.
Journal of cellular physiology, 234(10), 17727-17738 (2019-02-28)
This study aimed to investigate the mechanism of the ubiquitinase Aurora kinase A (AURKA) in the occurrence of osteoarthritis (OA) by mediating mitochondrial stress. Bioinformatic predictions revealed 2247 differentially expressed genes (DEGs) in the normal and OA tissues. According to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service