Skip to Content
Merck
All Photos(3)

Documents

HPA005128

Sigma-Aldrich

Anti-ENPEP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APA, Anti-Aminopeptidase A, Anti-CD249 antigen, Anti-Differentiation antigen gp160, Anti-EAP, Anti-Glutamyl aminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

SHLPSSTASPSGPPAQDQDICPASEDESGQWKNFRLPDFVNPVHYDLHVKPLLEEDTYTGTVSISINLSAPTRYLWLHLRETRITRLPELKRPSGDQVQVRRCFEYKKQEYVVVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENPEP(2028)

General description

ENPEP gene encodes the protein aminopeptidase A (APA), which is a membrane-bound ectopeptidase belonging to the zinc metalloprotease family. APA is expressed in a wide range of tissues, including the brain. Human ENPEP is localised to the chromosomal region 4q25. Human APA expressed in kidney is called gp160 and has a molecular weight of 160kDa.

Immunogen

Glutamyl aminopeptidase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

APA (aminopeptidase A) that is encoded by ENPEP gene cleaves acidic residues, such as aspartate and glutamate, from the N- terminal of its substrates and its activity is modulated by calcium levels. It is one of the key enzymes in the central renin-angiotensin system, and regulates hypertension. It converts angiotensin II to angiotensin III by cleaving the N-terminal aspartate residue. gp160 plays a key role in the development of kidney, and therefore is also known as the human kidney differentiation antigen. APA activity has been shown to be decreased in various renal cell carcinomas as compared to normal renal cells. This might lead to an increase in the level of angiotensin II in renal carcinomas, hence contributing to angiogenesis. It has been suggested that the formation of Amyloid β peptides is facilitated by APA, where it truncates the full length Amyloid β at the N-terminal.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85167

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena Blanco et al.
BMC cancer, 14, 386-386 (2014-06-03)
Advances in the knowledge of renal neoplasms have demonstrated the implication of several proteases in their genesis, growth and dissemination. Glutamyl-aminopeptidase (GAP) (EC. 3.4.11.7) is a zinc metallopeptidase with angiotensinase activity highly expressed in kidney tissues and its expression and
Yang Yang et al.
The Journal of biological chemistry, 288(35), 25638-25645 (2013-07-28)
Hypertension is regulated through both the central and systemic renin-angiotensin systems. In the central renin-angiotensin system, zinc-dependent aminopeptidase A (APA) up-regulates blood pressure by specifically cleaving the N-terminal aspartate, but not the adjacent arginine, from angiotensin II, a process facilitated
Stephen Tonna et al.
Gene, 410(1), 44-52 (2008-01-22)
The aminopeptidase A (APA) ectopeptidase is an integral membrane-bound zinc metalloprotease that cleaves aspartic and glutamic acidic residues from the N-terminus of a number of protein substrates that includes angiotensin II. Angiotensin II, the most vasoactive component of the renin-angiotensin-aldosterone
D M Nanus et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(15), 7069-7073 (1993-08-01)
gp160 is a cell surface differentiation-related glycoprotein of 160 kDa expressed by epithelial cells of the glomerulus and proximal tubule cells of the human nephron but only by a subset of renal cell carcinomas (RCCs). We have reported that gp160
Jean Sevalle et al.
Journal of neurochemistry, 109(1), 248-256 (2009-02-04)
Several lines of data previously indicated that N-terminally truncated forms of amyloid-beta (Abeta) peptides are likely the earliest and more abundant species immunohistochemically detectable in Alzheimer's disease-affected brains. It is noteworthy that the free N-terminal residue of full-length Abeta (fl-Abeta)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service