Skip to Content
Merck
All Photos(2)

Key Documents

HPA002852

Sigma-Aldrich

Anti-RPS6KA6 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-90 kDa ribosomal protein S6 kinase 6 antibody produced in rabbit, Anti-RSK-4 antibody produced in rabbit, Anti-Ribosomal S6 kinase 4 antibody produced in rabbit, Anti-Ribosomal protein S6 kinase α-6 antibody produced in rabbit, Anti-S6K-α 6 antibody produced in rabbit, Anti-p90-RSK 6 antibody produced in rabbit, Anti-pp90RSK4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PFKPASGKPDDTFCFDPEFTAKTPKDSPGLPASANAHQLFKGFSFVATSIAEEYKITPITSANVLPIVQINGNAAQFGEVYELKEDIGVGSYSVCKRCIHATTNMEFAVKIIDKSK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KA6(27330)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Immunogen

Ribosomal protein S6 kinase α-6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ribosomal protein S6 kinase, 90kDa, polypeptide 6 is a protein encoded by the RPS6KA6 gene in humans. It is a member of serine/threomine (Ser/Thr) kinase family and p53-related gene. The gene regulates the replicative and stress-induced senescence of different cells. It is a potential tumor suppressor in multiple cancers and may act as a valuable biomarker for the study of breast cancer carcinogenesis and progression. Its expression pattern and molecular mechanisms in RCCs (renal cell carcinoma) indicates that it could be a potential independent prognostic factor and serves as a new potential therapeutic target for RCC patients. Down-regulation of this gene in breast cancer suggests that it is a breast cancer suppressor gene. The gene is a putative tumor suppressor gene and is a target of the ERK signaling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74339

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuyun Li et al.
Medical oncology (Northwood, London, England), 31(1), 793-793 (2013-12-18)
Breast cancer is one of the most common cancers and is the second leading cause of cancer-related death in women worldwide. Ribosomal s6 kinase4 (RSK4) is a potential tumor suppressor in multiple cancers, while its role in breast cancer is
Jian-lun Liu et al.
Zhonghua zhong liu za zhi [Chinese journal of oncology], 33(6), 452-456 (2011-08-31)
To study the expression and clinical significance of ribosomal S6 kinase-4 (RSK-4) in breast cancer and explore the role of RSK-4 in the genesis and development of breast cancer. The expression levels of RSK-4 mRNA and protein were detected in
Summer B Dewdney et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(8), 2120-2129 (2011-03-05)
Effective treatments for advanced endometrial cancer are lacking. Novel therapies that target specific pathways hold promise for better treatment outcomes with less toxicity. Mutation activation of the FGFR2/RAS/ERK pathway is important in endometrial tumorigenesis. RPS6KA6 (RSK4) is a putative tumor
L Fan et al.
British journal of cancer, 109(5), 1137-1146 (2013-08-15)
The expression and function of ribosomal s6 protein kinase 4 (RSK4) in renal cell carcinoma (RCC) are unknown. Immunohistochemistry was used to detect the expression of RSK4 in RCC, and the relationship between RSK4 expression and clinicopathological features as well
Zhiyong Yin et al.
Cardiovascular diabetology, 11, 12-12 (2012-02-03)
The decrease and dysfunction of endothelial progenitor cells (EPCs) has been assumed as an important cause/consequence of diabetes mellitus (DM) and its complications, in which the senescence of EPCs induced by hyperglycemia may play an immensurable role. However, the mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service