Skip to Content
Merck
All Photos(6)

Key Documents

HPA002027

Sigma-Aldrich

Anti-MBL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MBP-C antibody produced in rabbit, Anti-MBP1 antibody produced in rabbit, Anti-Mannan-binding protein antibody produced in rabbit, Anti-Mannose-binding lectin antibody produced in rabbit, Anti-Mannose-binding protein C precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DGDSSLAASERKALQTEMARIKKWLTFSLGKQVGNKFFLTNGEIMTFEKVKALCVKFQASVATPRNAAENGAIQNLIKEEAFLGITDEKTEGQFVDLTGNRLTYTNWNEGEPNNAGSDEDCVLLLKNGQWNDVPCSTSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBL2(4153)

General description

MBL2 (mannose binding lectin 2) is a mannose-binding protein, expressed in neurons, astrocytes, microglia and belongs to the collectins family. It has also been found in oligodendrocytes of the frontal cortex of the HIV (human immunodeficiency virus)-1 infected brain.

Immunogen

Mannose-binding protein C precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-MBL2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MBL2 (mannose binding lectin 2) is associated with the activation of complement pathway, pathogen opsonization, chemotaxis, and activation of leukocytes and phagocytosis. Deficiency of MBL leads to to HIV (human immunodeficiency virus) transmission and disease progression. In bloodstream, it binds to the mannose residues or carbohydrates on pathogens such as bacteria, yeast, viruses, or parasites which helps to activate lectin complement pathway. With the help of two serine proteases, MBL-associated serine proteases (MASP-1 and MASP-2), MBL activates the complement factors C4 and C2 through a novel pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78225

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Fukami et al.
Muscle & nerve, 66(2), 175-182 (2022-05-19)
The mechanism of complement-mediated neurological injury in vasculitic neuropathy associated with systemic lupus erythematosus (SLE) and rheumatoid arthritis (RA) is unknown. The current study aimed to investigate the local activation of the complement system in vasculitic neuropathy associated with SLE
Jamie S Chua et al.
Journal of the American Society of Nephrology : JASN, 26(9), 2239-2247 (2015-01-13)
Complement activation has a major role in thrombotic microangiopathy (TMA), a disorder that can occur in a variety of clinical conditions. Promising results of recent trials with terminal complement-inhibiting drugs call for biomarkers identifying patients who might benefit from this
Jamie S Chua et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 32(8), 1147-1157 (2019-04-03)
Complement factor C4d was recently observed in renal biopsies from patients who had IgA nephropathy and a poor prognosis. We previously reported that C4d is a common denominator in microangiopathies. In this retrospective cohort study, we investigated whether C4d is
Kumud K Singh et al.
Neurobehavioral HIV medicine, 3, 41-52 (2011-08-20)
Mannose binding lectin (MBL) activates complement pathway that leads to pathogen opsonization and phagocytosis. MBL deficiency is linked to HIV transmission and disease progression. We sought to determine the role of MBL in HIV encephalitis (HIVE) by evaluating its presence
S Thiel et al.
Nature, 386(6624), 506-510 (1997-04-03)
The complement system comprises a complex array of enzymes and non-enzymatic proteins that is essential for the operation of the innate as well as the adaptive immune defence. The complement system can be activated in three ways: by the classical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service