Skip to Content
Merck
All Photos(1)

Key Documents

HPA001834

Sigma-Aldrich

Anti-CP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ceruloplasmin precursor antibody produced in rabbit, Anti-Ferroxidase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

YFSGNTYLWRGERRDTANLFPQTSLTLHMWPDTEGTFNVECLTTDHYTGGMKQKYTVNQCRRQSEDSTFYLGERTYYIAAVEVEWDYSPQREWEKELHHLQEQNVSNAFLDKGEFYIGSKYKKVVYRQYTDSTFRVPVERKAEEEHLGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CP(1356)

Immunogen

Ceruloplasmin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ceruloplasmin precursor (CP), a metalloprotein, is involved with the cellular iron export into plasma. Mutation in CP causes an inherited neurodegenerative disorder, aceruloplasminemia, which is an iron overload disease. In blood stream, Ferroportin (Fpn), a ferrous iron Fe(II) transporter helps in iron efflux. In absence of CP, iron remains bound to Fpn, unable to enter into the plasma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84476

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana De Domenico et al.
The EMBO journal, 26(12), 2823-2831 (2007-06-02)
Ferroportin (Fpn), a ferrous iron Fe(II) transporter responsible for the entry of iron into plasma, is regulated post-translationally through internalization and degradation following binding of the hormone hepcidin. Cellular iron export is impaired in mice and humans with aceruloplasminemia, an
Chin-Chung Shu et al.
Journal of clinical medicine, 8(5) (2019-05-28)
Increasing expression of programmed death-1 (PD-1) in patients with nontuberculous mycobacteria lung disease (NTM-LD) has been reported, but its role in clinical characteristics and outcomes remains unclear. We enrolled 96 participants, including 46 with Mycobacterium avium complex (MAC)-LD, 23 with
Rana Eltahan et al.
The Journal of eukaryotic microbiology, 66(3), 460-468 (2018-09-18)
Cryptosporidium parvum is one of the major species causing mild to severe cryptosporidiosis in humans and animals. We have previously observed that 2-deoxy-d-glucose (2DG) could inhibit both the enzyme activity of C. parvum hexokinase (CpHK) and the parasite growth in vitro. However
Susan C Fox et al.
Platelets, 30(3), 290-295 (2018-02-21)
Measurement of P-selectin on activated platelets as a means of measuring platelet function utilizing the technology described here has the advantage of not requiring immediate access to specialist equipment and expertise. Blood samples are activated, fixed, stored, and transported to
Juan Manuel Agraz-Cibrián et al.
Scandinavian journal of immunology, 88(5), e12714-e12714 (2018-09-19)
Liver cirrhosis (LC) is an inflammatory process associated with impaired functions in adaptive and innate immune responses at both systemic and local levels, also referred as Cirrhosis-Associated Immune Dysfunction. In this study, we evaluated the functionality of neutrophils from ascitic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service