Skip to Content
Merck
All Photos(3)

Documents

HPA001472

Sigma-Aldrich

Anti-CTCFL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Brother of the regulator of imprinted sites antibody produced in rabbit, Anti-CCCTC-binding factor antibody produced in rabbit, Anti-CTCF paralog antibody produced in rabbit, Anti-Transcriptional repressor CTCFL antibody produced in rabbit, Anti-Zinc finger protein CTCF-T antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

DGVCREKDHRSPSELEAQRTSGAFQDSVLEEEVELVLAPSEESEKYILTLQTVHFTSEAVELQDMSLLSIQQQEGVQVVVQQPGPGLLWLEEGPRQSLQQCVAISIQQELYSPQEMEVLQFHALEENVMVASEDSKLAVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTCFL(140690)

General description

CTCFL (CCCTC-binding factor (zinc finger protein)-like) gene encodes a protein that is expressed specifically by testis. It is a paralog of CTCF, a DNA binding protein, but varying in amino and carboxy termini. It is expressed in a mutually exclusive manner with CTCF during male germ cell development.

Immunogen

Transcriptional repressor CTCFL recombinant protein epitope signature tag (PrEST)

Application

Anti-CTCFL antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CTCFL (CCCTC-binding factor (zinc finger protein)-like) serves as a potential candidate for the epigenetic reprogramming factor in the male germ line. It is important in chromatin organization and gene expression. DNA methyltransferase 1 and 3B recruit CTCFL protein and regulate histone dimethylation, resulting in the control of BAG-1 (Bcl-2-binding protein) expression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79792

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Asano et al.
Oncotarget, 7(10), 11223-11237 (2016-02-06)
Cervical cancer is a major cause of cancer death in females worldwide. Cervical cancer stem-like cells (CSCs)/cancer-initiating cells (CICs) are resistant to conventional radiotherapy and chemotherapy, and CSCs/CICs are thought to be responsible for recurrence. Eradication of CSCs/CICs is thus
Marisol Salgado-Albarrán et al.
NPJ systems biology and applications, 8(1), 5-5 (2022-02-09)
High-grade serous ovarian carcinoma (HGSC) is the most lethal gynecologic malignancy due to the lack of reliable biomarkers, effective treatment, and chemoresistance. Improving the diagnosis and the development of targeted therapies is still needed. The molecular pathomechanisms driving HGSC progression
Marcelo Freitas et al.
Oncotarget, 4(4), 636-646 (2013-04-18)
Glioblastoma (GBM) confers a dismal prognosis despite advances in current therapy. Cancer-testis antigens (CTA) comprise families of tumor-associated antigens that are immunogenic in different cancers. The aim of this study was to determine the expression profile of a large number
William C Hines et al.
PloS one, 5(3), e9738-e9738 (2010-03-23)
BORIS (CTCFL) is the only known paralog of the versatile regulatory protein CTCF, a multifunctional DNA binding protein that mediates distinct gene regulatory functions involved in cell growth, differentiation, and apoptosis. Unlike CTCF, the expression of BORIS is normally restricted
Lunching Sun et al.
Cancer research, 68(8), 2726-2735 (2008-04-17)
In a previous genomic analysis, using somatic methyltransferase (DNMT) knockout cells, we showed that hypomethylation decreased the expression of as many genes as were observed to increase, suggesting a previously unknown mechanism for epigenetic regulation. To address this idea, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service