Skip to Content
Merck
All Photos(9)

Key Documents

HPA000798

Sigma-Aldrich

Anti-HSPA2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Heat shock 70 kDa protein 2 antibody produced in rabbit, Anti-Heat shock-related 70 kDa protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

RNAi knockdown
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ILNVTAADKSTGKENKITITNDKGRLSKDDIDRMVQEAERYKSEDEANRDRVAAKNALESYTYNIKQTVEDEKLRGKISEQDKNKILDKCQEVINWLDRNQMAEKDEYEHKQKELERVCNPIISKLYQGGPGGGSGGGGSGASGGPTIEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA2(3306)

General description

Heat shock-related 70kDa protein 2 (HSPA2) is encoded by the HSPA2 gene localized in the human chromosome 14q23.3. It is one of the isoforms of HSP-70 and is a cytosolic family member that undergoes methylation. HspA2 shows 85% homology to HspA1L isoform.

Immunogen

Heat shock-related 70 kDa protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-HSPA2 antibody produced in rabbit has also been used in:
  • duolink proximity ligation assay
  • immunoprecipitation
  • immunohistochemistry
  • immunocytochemistry
  • immunofluorescence microscopy
Anti-HSPA2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Heat shock-related 70kDa protein 2 is a testis-specific member of the HSP70 family and is correlated with sperm maturity, function and fertility. The protein is important for cancer cell growth and metastasis. It is identified as a potential cancer-promoting protein expressed at abnormal levels in a subset of cancers. HSPA2 has an important role in esophageal squamous cell carcinoma (ESCC) and overexpression of this gene may contribute to the malignant progression of ESCC. Its expression significantly correlates with sperm concentration and morphology. HSPA2 may play an important role in capacitation and fertilisation processes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70351

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinjie Bao et al.
Theranostics, 11(1), 132-146 (2021-01-05)
Rationale: The invasive behavior of non-functioning pituitary neuroendocrine tumors (NF-PitNEts) presents obstacles for complete surgical resection and is indicative of poor prognosis. Therefore, developing reliable diagnostic tools for identifying invasive PitNEts would be helpful in guiding surgical decisions and, in
Jürgen Radons
Cell stress & chaperones, 21(3), 379-404 (2016-02-13)
The 70-kDa heat shock protein (HSP70) family of molecular chaperones represents one of the most ubiquitous classes of chaperones and is highly conserved in all organisms. Members of the HSP70 family control all aspects of cellular proteostasis such as nascent
Hang Zhang et al.
World journal of surgical oncology, 11, 141-141 (2013-06-20)
Heat shock-related 70 kDa protein 2 (HSPA2) has been identified as a potential cancer-promoting protein expressed at abnormal levels in a subset of cancers. However, its important role in esophageal squamous cell carcinoma (ESCC) is hardly known by people. The
Dorota Scieglinska et al.
Anticancer research, 34(6), 2833-2840 (2014-06-13)
It has been shown that HSPA2 protein, a testis-enriched member of HSPA/HSP70 family, is important for cancer cell growth and metastasis. However, the status of HSPA2 expression in tumors and its clinical/prognostic significance are obscure. Herein we aimed to investigate
M Motiei et al.
Andrologia, 45(1), 66-72 (2012-06-08)
Heat-shock protein A2 (HSPA2) is a testis-specific member of the HSP70 family known to correlate with sperm maturity, function and fertility. The aim of this study was to compare expression of HSPA2 in fertile and infertile individuals using a recently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service