Skip to Content
Merck
All Photos(6)

Key Documents

H9664

Sigma-Aldrich

Anti-HMGB1 (HMG1) (N-terminal) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Amphoterin, Anti-High mobility group box 1, Anti-High mobility group protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IP
WB
clone:
polyclonal
species reactivity:
human, rat, mouse
citations:
13
technique(s):
immunoprecipitation (IP): 10 μg using HEK-293T cell lysates
indirect immunofluorescence: 1-2 μg/mL using paraformaldehyde-Triton fixed PC12 cultured cells.
indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using 3T3 cell lysates

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

species reactivity

human, rat, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 10 μg using HEK-293T cell lysates
indirect immunofluorescence: 1-2 μg/mL using paraformaldehyde-Triton fixed PC12 cultured cells.
indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using 3T3 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)
mouse ... Hmgb1(15289)
rat ... Hmgb1(25459)

General description

Anti-HMGB1 (HMG1) (N-terminal) is produced in rabbit using as immunogen a synthetic peptide corresponding to human HMGB1 conjugated to KLH. High Mobility Group B (HMGB) protein family includes HMGB1, HMGB2 and HMGB3, that are highly conserved and indistinguishable in their biochemical properties. HMGB1 is a 25 kDa protein of 215 amino acids and consists of two homologous HMG-boxes rich in basic amino acids, and an acidic tail at the carboxy-terminus.
HMGB proteins belong to the High Mobility Group (HMG) family of proteins that contain the HMG-box for binding and changing DNA structures .

Immunogen

synthetic peptide corresponding to amino acids 2-17 of human HMGB1, conjugated to KLH. The corresponding sequence is conserved in mouse and rat.

Application

Anti-HMGB1 (HMG1) (N-terminal) antibody is suitable for use in chemiluminescent immunoblot (using mouse heart homogenates) . The antibody can also be used in immunoprecipitation (10 μg using HEK-293T cell lysates), indirect immunofluorescence (1-2 μg/mL using paraformaldehyde-Triton fixed PC12 cultured cells), and western blot (1-2 μg/mL using 3T3 cell lysates).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

High Mobility Group B (HMGB-1 and 2) participate in the regulation of chromatin structure as well as being involved in transcription regulation, DNA repair, recombination, differentiation and extracellular signaling. It shows a defect in transcriptional enhancement of the glucocorticoid receptor, that indicates the important role for HMGB1 in proper transcriptional control by specific transcription factors. Increased expression of HMGB1 is observed in cancer cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Frescas et al.
Cell cycle (Georgetown, Tex.), 16(16), 1526-1533 (2017-06-27)
Age-related health decline has been attributed to the accumulation of senescent cells recognized in vivo by p16(Ink4a) expression. The pharmacological elimination of p16(Ink4a)-positive cells from the tissues of mice was shown to extend a healthy lifespan. Here, we describe a
High-mobility group box 1 and cancer
Tang D, et al.
Biochimica et Biophysica Acta - Gene Regulatory Mechanisms, 1799(1-2), 131-140 (2010)
Mao Wang et al.
American journal of respiratory cell and molecular biology, 52(2), 171-182 (2014-07-06)
The prolonged exposure to hyperoxia can compromise macrophage functions and contribute to the development of ventilator-associated pneumonia. High levels of extracellular high-mobility group box-1 (HMGB1) in the airways of mice exposed to hyperoxia can directly cause macrophage dysfunction. Hence, inhibition
Michal Stros
Biochimica et biophysica acta, 1799(1-2), 101-113 (2010-02-04)
HMGB proteins are members of the High Mobility Group (HMG) superfamily, possessing a unique DNA-binding domain, the HMG-box, which can bind non-B-type DNA structures (bent, kinked and unwound) with high affinity, and also distort DNA by bending/looping and unwinding. HMGBs
Xun Li et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 51(5), 2198-2211 (2018-12-12)
Our previous study demonstrated that a deficiency of microRNA 21 (miR-21) protects mice from acute pancreatitis, yet the underlying molecular networks associated with miR-21 in pancreatitis and pancreatitis-associated lung injury remain unexplored. We used next generation sequencing to analyze gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service