Skip to Content
Merck
All Photos(1)

Key Documents

H8666

Sigma-Aldrich

Transforming Growth Factor-β2 human

≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

TGF-β2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Transforming Growth Factor-β2 human, TGF-β2, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.5 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 25 kDa (non-glycosylated)

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TGFB2(7042)

Looking for similar products? Visit Product Comparison Guide

General description

Transforming growth factor-β2 (TGF-β2) belongs to the TGF-β superfamily. All threes isoforms (TGF-β1, TGF-β2 and TGF-β3) are known to share similar structure and function. The TGFB2 gene is mapped to human chromosome 1q41 and is localized to extracellular matrix. The active form of TGF-β precursor is produced upon proteolytic cleavage. TGF-β1 and TGF-β2 have 70% amino acid sequence identity.

Biochem/physiol Actions

TGF-β2 is important for immune homeostasis by balancing lymphocyte proliferation, apoptosis, hematopoiesis, and embryogenesis. TGF-β2 is crucial in cell growth, differentiation, and survival. TGF-β2 is a strong growth inhibitor for normal and transformed epithelial, lymphoid, fibroblast, and keratinocyte cells. TGF-β is associated with the process of tumor development, progression and metastasis. TGF-β2 is a tumor suppressor in the early stages of carcinogenesis, but in the later stages acts as a tumor promoter by inducing epithelial-mesenchymal transition and stimulating angiogenesis. TGF-β2 inhibits antitumor action of NK (natural killer) cells, T-cells, macrophages, monocytes and neutrophils. TGF-β is also associated with inflammation and wound healing. The TGF-β gene is overexpressed in glioma tumors and also in colon cancer, gastric cancer and cervical lesions.
Transforming growth factor-β2 (TGF-β2), like TGF-β1, is produced by many cell types and reported to be most concentrated in mammalian platelets.

Preparation Note

HumanKine TGF-β2 is expressed in human HEK 293 cells as a mature, non-glycosylated, disulfide-linked homodimer with a predicted molecular mass of approx. 25 kDa.

Analysis Note

The specific activity was determined by the dose-dependent inhibition of IL-4 induced proliferation of mouse HT-2 (BALB/c spleen activated by sheep erythrocytes in the presence of IL-2).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of zinc finger E-box binding homeobox 1 and transforming growth factor-β2 expression in bladder cancer tissue in comparison with healthy adjacent tissue.
Mahdavinezhad A, et al.
Investigative and Clinical Urology, 58(2), 140-145 (2017)
A Signaling Pathway Involving TGF-β2 and Snail in Hair Follicle Morphogenesis
Jamora C, et al.
PLoS Biology, 3.1 (2004)
Sunder Sims-Lucas et al.
Differentiation; research in biological diversity, 79(4-5), 272-284 (2010-02-19)
Many members of the transforming growth factor-beta (TGF-beta) superfamily have been shown to be important regulators of metanephric development. In this study, we characterized the effect of TGF-beta2 on metanephric development. Rat and mouse metanephroi cultured in the presence of
Loss-of-function mutations in TGFB2 cause a syndromic presentation of thoracic aortic aneurysm
Lindsay ME, et al.
Nature Genetics, 44(8), 922?927-922?927 (2012)
Curcumin suppresses doxorubicin-induced epithelial?mesenchymal transition via the inhibition of TGF-β and PI3K/AKT signaling pathways in triple-negative breast cancer cells.
Chen WC, et al.
Journal of Agricultural and Food Chemistry, 61.48, 11817-11824 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service