Skip to Content
Merck
All Photos(1)

Key Documents

H6875

Sigma-Aldrich

Hippuryl-L-phenylalanine

Synonym(s):

Hippuryl-Phe, N-Benzoyl-Gly-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O4
CAS Number:
Molecular Weight:
326.35
Beilstein:
2167818
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

acetic acid: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)c2ccccc2

InChI

1S/C18H18N2O4/c21-16(12-19-17(22)14-9-5-2-6-10-14)20-15(18(23)24)11-13-7-3-1-4-8-13/h1-10,15H,11-12H2,(H,19,22)(H,20,21)(H,23,24)/t15-/m0/s1

InChI key

CCLJGZGVIQBNDH-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hippuryl-L-phenylalanine has been used as a substrate for screening carboxypeptidase activity in Trogoderma granarium, Bactrocera oleae Gmelin and Apodiphus amygdali.

Biochem/physiol Actions

Hippuryl-L-phenylalanine is a substrate for carboxypeptidase A enzyme.

Substrates

Substrate for carboxypeptidase A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of kininase I and kininase II activities in human urine by high-performance liquid chromatography.
G Porcelli et al.
Journal of chromatography, 414(2), 423-428 (1987-03-06)
E Normant et al.
Neuropeptides, 30(1), 13-17 (1996-02-01)
We have designed two radioactive substrates, hippuryl-L-[3H]phenylalanine and 3-(p-hydroxy, m-[125I]phenyl)propionic acid ([125I]Bolton reagent) derivative of L-arginyl-L-phenylalanine, i.e. [125I]BRF, for a highly sensitive assay of carboxypeptidase A (CPA) activity. After cleavage of the C-terminal phenylalanine residue by CPA, the radioactive product
Anke Schwarzenberger et al.
Frontiers in microbiology, 11, 586120-586120 (2020-11-17)
The harmful bloom-forming cyanobacterium Planktothrix is commonly considered to be nutritionally inadequate for zooplankton grazers, resulting in limited top-down control. However, interactions between Planktothrix and zooplankton grazers are poorly understood. The food quality of Planktothrix is potentially constrained by morphological
J F Sebastian et al.
Canadian journal of biochemistry, 56(5), 329-333 (1978-05-01)
3,3-Diphenylpropanoate (DPP) activates the carboxypeptidase A catalyzed hydrolysis of benzoylglycyl-L-phenylalanine (BzGly-L-Phe) (Ka = 2.1 x 10 (-3) M) and inhibits ester hydrolysis uncompetitively (K1 =2.1 X 10 (-3) M). A common modifier binding site located adjacent to the peptide and
Product stabilization of the two forms of carboxypeptidase A from buffalo pancreas
Sud M and Dua RD
Journal of Biosciences, 9(1-2), 83-90 (1985)

Protocols

Carboxypeptidase A activity measured via continuous spectrophotometric rate determination assay with hippuryl-L-phenylalanine substrate.

Carboxypeptidase A activity measured via continuous spectrophotometric rate determination assay with hippuryl-L-phenylalanine substrate.

Carboxypeptidase A activity measured via continuous spectrophotometric rate determination assay with hippuryl-L-phenylalanine substrate.

Carboxypeptidase A activity measured via continuous spectrophotometric rate determination assay with hippuryl-L-phenylalanine substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service