Skip to Content
Merck
All Photos(6)

Key Documents

H5147

Sigma-Aldrich

Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse

clone BRM-22, ascites fluid

Synonym(s):

HSP70 Antibody - Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse, Hsp70 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BRM-22, monoclonal

mol wt

antigen 70 kDa

species reactivity

bovine, chicken, guinea pig, plant, Drosophila, rat, human, nematode, rabbit, hamster

technique(s)

electron microscopy: suitable
immunohistochemistry (frozen sections): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:5,000 using bovine brain extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

A variety of environmental disruptions, such as a sudden increase in temperature, induce cells to rapidly synthesize a group of polypeptides known as heat shock (stress) proteins. Eukaryotic cells contain a multigene family that encodes several closely related 70 kD stress proteins (the HSP70 family) that differ in their intracellular location and regulation.These include four proteins: the constitutive (or cognate) HSP73, the stress-inducible HSP72 and the glucose regulated proteins grp78 (or BiP) and grp75.
Monoclonal Anti-Heat Shock Protein 70 (mouse IgG1 isotype) is derived from the BRM-22 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with purified bovine brain HSP70. Heat shock protein family A (Hsp70) member 1A (HSPA1A), is encoded by the gene mapped to human chromosome 6p21.33. It is widely expressed in all tissues. The encoded protein is characterized with an N-terminal ATPase domain of 45 kDa and a C-terminal substrate binding domain of 25 kDa.

Specificity

By immunoblotting, the antibody localizes both the constitutive (HSP73) and inducible (HSP72) forms of HSP70. The antibody recognizes HSP70 from brain and other tissue. Immunofluorescent staining demonstrates a rapid and reversible accumulation of the HSP70 protein within the nucleus of heat-shocked (42 °C, 1 hour) human fibroblasts.

Immunogen

HSP70 isolated from bovine brain

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
Western Blotting (4 papers)
Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblot
  • dot blot
  • immunocytochemistry

The antibody titer of at least 1:5,000 was determined by immunoblotting using bovine brain extract.

Biochem/physiol Actions

Members of the HSP70 family play a major role in the folding, unfolding and translocation of polypeptides as well as in the assembly and disassembly of oligomeric protein complexes. In addition, several possible roles have been attributed to the HSP70 family of proteins, in the immune response. It has been shown that alcoholic liver disease is associated with intracytoplasmic accumulation of HSP70. HSP72 was found to increase dramatically in the brains of Alzheimer′s disease patient, and was localized exclusively in neuritic plaques and neurofibrillary tangles. HSP70 concentrates in nuclei during heat shock and returns to the cytoplasm when the shock is removed.

Other Notes

This product can be found as purified product that was produced using cell culture hybridoma product.
SAB4200714 Anti-Heat Shock Protein 70 (HSP70) antibody, Mouse monoclonal
clone BRM-22, purified from hybridoma cell culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Xie et al.
Cell stress & chaperones, 21(5), 907-914 (2016-07-21)
Although accumulating evidence indicates that heat shock protein 70 (HSP70) could be secreted into plasma and its levels have been found to have an ambiguous association with atherosclerosis, our knowledge for the exact role of circulating HSP70 in the development
Anne Lyytinen et al.
PloS one, 7(2), e31446-e31446 (2012-02-10)
Individuals of widely spread species are expected to show local adaption in temperature tolerance as they encounter a range of thermal conditions. We tracked thermal adaptations of the Colorado potato beetle (Leptinotarsa decemlineata) that invaded Europe within the last 100
Mohan E Tulapurkar et al.
PloS one, 10(2), e0118010-e0118010 (2015-02-07)
Sepsis, a devastating and often lethal complication of severe infection, is characterized by fever and dysregulated inflammation. While infections activate the inflammatory response in part through Toll-like receptors (TLRs), fever can partially activate the heat shock response with generation of
hsp70 genes in the human genome: Conservation and differentiation patterns predict a wide array of overlapping and specialized functions
Brocchieri L, et al.
BMC Evolutionary Biology, 8(1), 1-20 (2008)
Bacterial lipopolysaccharide augments febrile-range hyperthermia-induced heat shock protein 70 expression and extracellular release in human THP1 cells
Tulapurkar ME, et al.
Testing, 10(2), e0118010-e0118010 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service