Skip to Content
Merck
All Photos(2)

Key Documents

H4036

SAFC

L-Histidine monohydrochloride monohydrate

Pharma Manufacturing

Synonym(s):

L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.26

biological source

non-animal source

Quality Level

Assay

≥98.5%

form

powder

manufacturer/tradename

Ajinomoto

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, heavy metals, residual solvents, tested

mp

254 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

application(s)

pharmaceutical (small molecule)

foreign activity

cytotoxicity, tested

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.

Application

L-Histidine, an essential amino acid. It is used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-Histidine monohydrochloride monohydrate is a modified version of L-Histidine with improved solubility.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bakul Bhatnagar et al.
The journal of physical chemistry. B, 123(27), 5690-5699 (2019-07-02)
Protein/ice interactions are investigated by a novel method based on measuring the characteristic features of X-ray diffraction (XRD) patterns of hexagonal ice (Ih). Aqueous solutions of four proteins and other solutes are studied using high-resolution synchrotron XRD. Two pharmaceutical proteins
Martin Maldonado et al.
Neurochemistry international, 85-86, 31-39 (2015-05-06)
The effect of reserpine on histamine (HA) and tele-methylhistamine (N(τ)-MHA) in hypothalamus and cortex of rats was analyzed and compared to catecholamines. IP injection of reserpine (5 mg/kg) confirmed the effectiveness of reserpine treatment on noradrenaline and dopamine levels. Our in-vitro
Verena Saller et al.
Journal of pharmaceutical sciences, 104(4), 1440-1450 (2015-01-22)
In a typical manufacturing setup for biopharmaceutical drug products, the fill and dosing pump is placed after the final sterile filtration unit in order to ensure adequate dispensing accuracy and avoid backpressure peaks. Given the sensitivity of protein molecules, peristaltic
Aldo Roda et al.
The Analyst, 139(24), 6494-6501 (2014-10-25)
Increasingly, smartphones are used as portable personal computers, revolutionizing communication styles and entire lifestyles. Using 3D-printing technology we have made a disposable minicartridge that can be easily prototyped to turn any kind of smartphone or tablet into a portable luminometer
Kristin Bjornsdottir-Butler et al.
Journal of food protection, 78(7), 1335-1342 (2015-07-22)
Recent developments in detection and enumeration of histamine-producing bacteria (HPB) have created powerful molecular-based tools to better understand the presence of spoilage bacteria and conditions, resulting in increased risk of scombrotoxin fish poisoning. We examined 235 scombrotoxin-forming fish from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service