Skip to Content
Merck
All Photos(1)

Documents

H2898

Sigma-Aldrich

6β-Hydroxytestosterone

≥97% (HPLC)

Synonym(s):

4-Androstene-6β,17β-diol-3-one, 6β, 17β,-Dihydroxyandrost-4-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

methanol: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](C[C@@H](O)C4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,16-17,21-22H,3-8,10H2,1-2H3/t12-,13-,14-,16+,17-,18+,19-/m0/s1

InChI key

XSEGWEUVSZRCBC-ZVBLRVHNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6β-Hydroxytestosterone has been used as a standard in high-performance liquid chromatography (HPLC) analysis to measure testosterone 6 β -hydroxylase activity in rat liver microsomes, and measure cytochrome P450 3A (CYP3A) activity in the intestinal spheroid model.

Biochem/physiol Actions

6β-Hydroxytestosterone is a metabolite generated by brain testosterone cytochrome P450 1B1 (CYP1B1). It may elicit angiotensin II-induced neurogenic hypertension and inflammation in male mice.
6β-Hydroxytestosterone is a CYP3A4 metabolite; androgenic.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purnima Singh et al.
Hypertension (Dallas, Tex. : 1979), 76(3), 1006-1018 (2020-08-07)
Previously, we showed that peripheral administration of 6β-hydroxytestosterone, a CYP1B1 (cytochrome P450 1B1)-generated metabolite of testosterone, promotes angiotensin II-induced hypertension in male mice. However, the site of action and the underlying mechanism by which 6β-hydroxytestosterone contributes to angiotensin II-induced hypertension
Laura M Langan et al.
Frontiers in pharmacology, 9, 947-947 (2018-09-07)
Compared to two-dimensional (2D) cell culture, cellular aggregates or spheroids (3D) offer a more appropriate alternative in vitro system where individual cell-cell communication and micro-environment more closely represent the in vivo organ; yet we understand little of the physiological conditions
Vilasinee Hirunpanich et al.
Drug metabolism and disposition: the biological fate of chemicals, 34(2), 305-310 (2005-11-22)
To investigate the pharmacokinetic interaction between cyclosporin A (CsA) and docosahexaenoic acid (DHA) in vivo, 5 mg/kg CsA was orally or intravenously coadministered with DHA (50-200 microg/kg) to rats. The effect of DHA on CYP3A activity was determined using rat
Takashi Furuta et al.
Steroids, 68(7-8), 693-703 (2003-09-06)
A method is described for the preparation of two types of multi-labeled 6 beta-hydroxycortisol containing either five deuterium atoms at C-19 methyl and C-1 methylene or four 13C atoms at C-1, C-2, C-4, and C-19 in addition to the five
Ying-Hong Wang et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(5), 664-671 (2005-02-04)
The genetic basis for polymorphic expression of CYP3A5 has been recently identified, but the significance of CYP3A5 expression is unclear. The purpose of this study is to quantify the capability of verapamil, a mechanism-based inhibitor of CYP3A, and its metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service