Skip to Content
Merck
All Photos(2)

Key Documents

H2662

Sigma-Aldrich

Anti-Histone Deacetylase 7 (HDAC7) (KG-17) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HD7, Anti-HD7A, Anti-HDAC7A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~105 kDa

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): 1.0-1.5 μg using RIPA extract of 293T cells expressing recombinant human HDAC7
indirect immunofluorescence: 1:50 using rat NRK cells
western blot: 1:1,000 using whole extracts of mouse NIH-3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC7(51564)
mouse ... Hdac7(56233)
rat ... Hdac7a(84582)

General description

Mammalian histone deacetylases (HDACs) can be divided into three classes according to sequence homology. Class I consists of the yeast transcriptional regulatory protein Rpd3-like proteins HDAC1, HDAC2, HDAC3, and HDAC8. Class II consists of the yeast Hda1-like proteins HDAC4, HDAC5, HDAC6, HDAC7, HDAC9 and HDAC10. Class III consists of the yeast sirtuin 2 (Sir2)-like proteins. Although HDAC7 is localized mostly to the cell nucleus, it is also found in the cytoplasm.

Specificity

Anti-Histone Deacetylase 7 recognizes human, mouse, and rat HDAC7.

Immunogen

Synthetic peptide corresponding to amino acids of human HDAC7, conjugated to KLH.

Application

Anti-Histone Deacetylase 7 (HDAC7) (KG-17) antibody produced in rabbit has been used in
  • indirect immunofluorescene
  • immunoblotting
  • immunoprecipitation

Biochem/physiol Actions

Class II histone deacetylases (HDACs) have been implicated in the regulation of muscle differentiation. Interaction of HDAC4, -5, and -7 with members of the myocyte enhancer factor-2 (Mef2) family of transcription factors represses their transcriptional activity and prevents myogenesis. Shuttling of HDAC7 between the cell nucleus and the cytoplasm is controlled by a mechanism involving calmodulin independent kinase I (CaMKI) and 14-3-3 proteins. The HDAC7 enzymatic activity depends on its interaction with the class I HDAC3, and the corepressors silencing mediator of retinoic acid and thyroid hormone receptors (SMRT) and nuclear receptor co-repressor (N-COR). HDAC7 also interacts with the transcriptional repressor B-cell lymphoma 6 (BCL-6).

Physical form

Solution in 0.01 M phosphate buffered saline containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erasers of histone acetylation: the histone deacetylase enzymes
Seto E and Yoshida M
Cold Spring Harbor Perspectives in Biology, 6(4), a018713-a018713 (2014)
Splicing of HDAC7 modulates the SRF-myocardin complex during stem-cell differentiation towards smooth muscle cells
Margariti A, et al.
Journal of Cell Science, 122(4), 460-470 (2009)
Carina Mello Guimaraes Meyers et al.
Bone, 159, 116393-116393 (2022-03-24)
Protein kinase D (PRKD) family kinases are required for formation and function of osteoclasts. However, the substrates of PRKD in osteoclasts are unknown. To identify PRKD-dependent protein phosphorylation in osteoclasts, we performed a quantitative LC-MS/MS phosphoproteomics screen for proteins showing
Human HDAC7 Histone Deacetylase Activity Is Associated with HDAC3in Vivo
Fischle W, et al.
Test, 276(38), 35826-35835 (2001)
Class II histone deacetylases are directly recruited by BCL6 transcriptional repressor
Lemercier C, et al.
Test, 277(24), 22045-22052 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service