Skip to Content
Merck
All Photos(1)

Documents

H0388

Sigma-Aldrich

Hexacosanoic acid

≥95% (capillary GC)

Synonym(s):

Cerotic acid, Cerotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)24COOH
CAS Number:
Molecular Weight:
396.69
Beilstein:
1799681
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (capillary GC)

form

powder

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)

InChI key

XMHIUKTWLZUKEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Aubourg et al.
The New England journal of medicine, 329(11), 745-752 (1993-09-09)
Adrenomyeloneuropathy is an X-linked recessive disorder characterized by myelopathy, peripheral neuropathy, and cerebral demyelination, which develop in association with the accumulation of very-long-chain fatty acids. The administration of oleic and erucic acids inhibits the synthesis of very-long-chain fatty acids. Recently
Pedro Brites et al.
Brain : a journal of neurology, 132(Pt 2), 482-492 (2008-11-22)
Peroxisomes are organelles responsible for multiple metabolic pathways including, the biosynthesis of plasmalogens, a class of phospholipids, and the beta-oxidation of very-long-chain fatty acids (VLCFA). Lack of peroxisomes or dysfunction in any of their normal functions is the cellular basis
Marc Engelen et al.
Journal of inherited metabolic disease, 35(6), 1137-1145 (2012-03-27)
X-linked adrenoleukodystrophy (X-ALD) is caused by mutations in the ABCD1 gene encoding ALDP, an ATP-binding-cassette (ABC) transporter located in the peroxisomal membrane. ALDP deficiency results in impaired peroxisomal β-oxidation and the subsequent accumulation of very long-chain fatty acids (VLCFA; >
Hugo W Moser et al.
Archives of neurology, 62(7), 1073-1080 (2005-07-13)
To identify asymptomatic boys with X-linked adrenoleukodystrophy who have a normal magnetic resonance image (MRI), and to assess the effect of 4:1 glyceryl trioleate-glyceryl trierucate (Lorenzo's oil) on disease progression. Eighty-nine boys (mean +/- SD baseline age, 4.7 +/- 4.1
A Sadeghi-Nejad et al.
The New England journal of medicine, 322(1), 13-16 (1990-01-04)
Adrenoleukodystrophy, a sex-linked peroxisomal disorder that results in the impaired oxidation of long-chain saturated fatty acids and causes neurologic impairment, is a rare cause of Addison's disease in children. Adrenomyeloneuropathy is the name given to a biochemically identical but milder

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service