Skip to Content
Merck
All Photos(1)

Key Documents

GW20056A

Sigma-Aldrich

Anti-Transcription Factor E3 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TFE3, Anti-Transcription factor binding to IGHM enhancer 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-20056A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFE3(7030)

Related Categories

Immunogen

Immunogen Sequence: GI # 8659574, sequence 432-541
Recombinant transcription factor binding to IGHM enhancer 3

Application

Anti-Transcription Factor E3 antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for cell staining at a working dilution of 1:200.

Biochem/physiol Actions

TFE3 (Transcription factor E3) gene encodes a helix-loop-helix transcription factor that contains a helix-loop-helix domain and binds to MUE3-type E-box sequences (5′-CANNTG-3′). It plays a key role in transforming growth factor β (TGF-β) induced activation of the plasminogen activator inhibitor-1 (PAI-1) promoter. Chromosomal translocation resulting in the fusion of TFE3 gene to a novel gene, designated PRCC, is observed in papillary renal cell carcinomas. A chromosomal aberration in TFE3 is observed in patients with alveolar soft part sarcoma.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Clark et al.
Oncogene, 15(18), 2233-2239 (1997-12-11)
We demonstrate that the cytogenetically defined translocation t(X;1)(p11.2;p34) observed in papillary renal cell carcinomas results in the fusion of the splicing factor gene PSF located at 1p34 to the TFE3 helix-loop-helix transcription factor gene at Xp11.2. In addition we define
P Argani et al.
The American journal of pathology, 159(1), 179-192 (2001-07-05)
The unbalanced translocation, der(17)t(X;17)(p11.2;q25), is characteristic of alveolar soft part sarcoma (ASPS). We have recently shown that this translocation fuses the TFE3 transcription factor gene at Xp11.2 to ASPL, a novel gene at 17q25. We describe herein eight morphologically distinctive
H Beckmann et al.
Genes & development, 4(2), 167-179 (1990-02-01)
The muE3 motif within the immunoglobulin heavy-chain enhancer is required for full enhancer activity and is known to bind one, or perhaps a family, of related ubiquitous nuclear proteins. Here, we present the isolation of a cDNA that encodes an
M A Weterman et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(26), 15294-15298 (1996-12-24)
The (X;1)(p11;q21) translocation is a recurrent chromosomal abnormality in a subset of human papillary renal cell carcinomas, and is sometimes the sole cytogenetic abnormality present. Via positional cloning, we were able to identify the genes involved. The translocation results in
X Hua et al.
Genes & development, 12(19), 3084-3095 (1998-10-09)
Members of the TGF-beta superfamily influence a broad range of biological activities including stimulation of wound healing and inhibition of cell growth. TGF-beta signals through type I and II receptor serine/ threonine kinases and induces transcription of many genes including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service