Skip to Content
Merck
All Photos(1)

Key Documents

GERPN850E

ECL Plex Fluorescent Rainbow Markers

Cytiva RPN850E, pack of 120 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.31
Pricing and availability is not currently available.

feature

wetted part: no

packaging

pack of 120 μL

manufacturer/tradename

Cytiva RPN850E

color

Brownish-red

solubility

cold water: soluble (Soluble in cold water and hot water)

shipped in

dry ice

storage temp.

−20°C

General description

ECL Plex Fluorescent Rainbow Markers are protein molecular weight markers, optimized for use with the ECL Plex Western blotting detection system.
They are optimized to provide visible marker bands on gels and membranes, as well as fluorescent images using Cy3 and Cy5 channels.

Application

ECL Plex Fluorescent Rainbow Markers has been used for quantitative western blots.[1][2]

Features and Benefits

  • Visible marker bands on gels and blots
  • Fluorescent images in both Cy3 and Cy5 channels

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL Plex is a trademark of Cytiva
Rainbow is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of cell uptake pathways for tumor inhibitor lysyl oxidase propeptide.
Ozdener G B, et al.
Molecular Oncology, 10(1), 1-23 (2016)
Expression of xenobiotic transporters in the human renal proximal tubule cell line RPTEC/TERT1.
Aschauer L, et al.
Toxicology in vitro, 30(1), 95-105 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service