Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9180-06

PD MultiTrap G-25

Cytiva 28-9180-06

Synonym(s):

G-25 MultiTrap, MultiTrap G-25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

material

polypropylene and polyethylene

sterility

non-sterile

feature

wetted part: no

packaging

pkg of 4 ea

manufacturer/tradename

Cytiva 28-9180-06

storage condition

dry media

size

127.8 mm × 85.5 mm × 30.6 mm

working volume

70—130 μL

matrix

cross-linked dextran

particle size

>50 μm (dry)
85-260 μm

pore size

MWCO

cleaning

2-13

working range

2-13

suitability

suitable for bioprocess medium

General description

PD MultiTrap G-25 is designed for high-throughput desalting and buffer exchange of multiple samples. Supplied in packs of 4 plates. Instructions included. Plates can be processed by vacuum or centrifugation. PD MultiTrap G-25 is prepacked with 500 μ Sephadex® G-25 Medium/well. The 96-well plates are manufactured from biocompatible polypropylene. High desalting capacity (>85%) and recovery (70-90%).

Application

Techniques & Applications: DNA and Protein sample preparation. Desalting/Buffer Exchange/Cleanup.

Features and Benefits

  • Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with high reproducibility well-to-well and plate-to-plate.
  • Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds.
  • High desalting capacity.
  • Suitable for manual use as well as for automation together with robotic systems.
  • Sample preparation in the range of 70 to 130 μl without sample dilution.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)

Legal Information

MultiTrap is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

Protocols

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service