Key Documents
GE18-3216-03
Labmate PD-10 Buffer Reservoir
Cytiva 18-3216-03, pack of 1 ea
About This Item
UNSPSC Code:
41115700
NACRES:
NA.56
sterility
non-sterile
feature
wetted part: no
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 18-3216-03
General description
LabMate PD-10 buffer reservoir.
Features and Benefits
  • Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex® G-25 Medium.
  • LabMate Buffer Reservoir makes it easier and more convenient to equilibrate PD-10 columns.
  • Easy to use and equilibrate; apply sample; elute.
Legal Information
Sephadex is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
This page describes characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (high sub) media and columns from Cytiva.
Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.
Protocols
This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.
This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.
This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.
This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.
See All