Skip to Content
Merck
All Photos(2)

Key Documents

GE17-5143-01

HiTrap® Benzamidine Fast Flow

Cytiva 17-5143-01, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

p-aminobenzamidine

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-5143-01

storage condition

(0.05 M Sodium Acetate, pH 4.0 in 20% Ethanol)

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning

1-9

working range

2-8

storage temp.

2-8°C

General description

HiTrap™ Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose™ 4 Fast FLow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.

Application

HiTrap™ Benzamidine FF (high sub) are prepacked 1 mL and 5 mL columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein. Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates.

The medium, Benzamidine Sepharose™ 4 Fast FLow (high sub), is also available as lab packages and is an excellent choice for scale-up. The columns can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTAdesign or FPLC™ System.

Features and Benefits

  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant Proteins.
  • Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates.
  • HiTrap™ Benzamidine FF for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
  • High binding capacity.
  • The columns can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA™ design or FPLC™ System.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

This page shows troubleshooting strategies for cleavage methods using Cytiva products.

This page shows how to cleave and purify GST-tagged proteins bound to Glutathione Sepharose in batch mode from Cytiva. Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, and Glutathione Sepharose 4B can all be used for cleavage and purification of GST-tagged proteins in batch.

Protocols

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service