Key Documents
GE10600122
Amersham Hybond® P Western blotting membrane sandwich, PVDF
pore size 0.2 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600122
sheet W × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
pore size
0.2 μm pore size
Related Categories
General description
Amersham Hybond P PVDF blotting membrane, with 0.2 µm pore size, are designed for use with chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000.
Features and Benefits
  • Contains 10 Western blotting sandwiches, consisting of precut Amersham Hybond P 0.2 µm PVDF membranes preassembled with 2 × 3MM Chr Blotting papers.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Guoxia Zhu et al.
Neural plasticity, 2020, 8823785-8823785 (2020-10-22)
Sound conditioning (SC) is defined as "toughening" to lower levels of sound over time, which reduces a subsequent noise-induced threshold shift. Although the protective effect of SC in mammals is generally understood, the exact mechanisms involved have not yet been
Articles
This page describes troubleshooting strategies in the detection of GST-tagged proteins.
Protocols
This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.