GE10600079
Amersham™ Protran® Premium Western blotting membranes, nitrocellulose
pore size 0.45 μm, sheet W × L 100 mm × 100 mm, pkg of 10 ea
About This Item
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600079
sheet W × L
100 mm × 100 mm
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- High binding capacity (162 – 180 µg IgG/cm2).
- Suitable for proteins of a wide range of molecular weights.
- Unsupported, 100% pure nitrocellulose membrane for use with all ECL Western blotting systems and protein blotting applications.
- Available in popular precut sizes for laboratory application.
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service