Skip to Content
Merck
All Photos(1)

Key Documents

G6751

Sigma-Aldrich

α-Glycerophosphate Dehydrogenase from rabbit muscle

Type I, ammonium sulfate suspension, 100-300 units/mg protein

Synonym(s):

sn-Glycerol-3-phosphate Dehydrogenase from rabbit muscle, sn-Glycerol-3-phosphate:NAD+ 2-oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

rabbit muscle

Quality Level

type

Type I

form

ammonium sulfate suspension

specific activity

100-300 units/mg protein

storage condition

(Tightly closed)

technique(s)

activity assay: suitable

color

white

foreign activity

Lactic dehydrogenase, pyruvate kinase, aldolase, and glyceraldehyde-3-phosphate dehydrogenase ≤0.01%
Triosephosphate isomerase ≤0.02%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

α-Glycerophosphate dehydrogenase is a membrane protein and a flavin-linked primary dehydrogenase.

Research area: Cell Signaling

Application

α-Glycerophosphate Dehydrogenase from rabbit muscle has been used:
  • in the reaction mixture to measure the glycerol kinase activity
  • to demonstrate compartmentalized enzymatic reactions where NADH is involved
  • in the reaction mixture assay for L-fuculose-1-phosphate aldolase

Biochem/physiol Actions

α-Glycerophosphate dehydrogenase catalyzes the oxidation of L-α-glycerophosphate (Glp) to form dihydroxyacetone phosphate (DHAP) and hydrogen peroxide (H2O2). This enzyme is associated with respiratory electron transport chain, glycolysis, and phospholipid metabolism.

Unit Definition

One unit will convert 1.0 μmole of dihydroxyacetone phosphate to α-glycerophosphate per min at pH 7.4 at 25 °C.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4 and 0.1 g/L EDTA solution, pH 6.0

Preparation Note

Prepared by modification of the method of Fondy.

Analysis Note

Protein determined by biuret.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimal conditions for biomass and recombinant glycerol kinase production using the yeast Pichia pastoris
Aizemberg R, et al.
Food Technology and Biotechnology, 49(3), 329-329 (2011)
Structural studies on nicotinamide adenine dinucleotide-linked L-glycerol 3-phosphate dehydrogenase crystallized from rat skeletal muscle.
T P Fondy et al.
The Journal of biological chemistry, 243(11), 3148-3160 (1968-06-10)
Joanne I Yeh et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(9), 3280-3285 (2008-02-26)
Sn-glycerol-3-phosphate dehydrogenase (GlpD) is an essential membrane enzyme, functioning at the central junction of respiration, glycolysis, and phospholipid biosynthesis. Its critical role is indicated by the multitiered regulatory mechanisms that stringently controls its expression and function. Once expressed, GlpD activity
Somchart Maenpuen et al.
The FEBS journal, 282(16), 3043-3059 (2015-02-26)
L-α-glycerophosphate oxidase is an FAD-dependent enzyme that catalyzes the oxidation of L-α-glycerophosphate (Glp) by molecular oxygen to generate dihydroxyacetone phosphate (DHAP) and hydrogen peroxide (H2O2). The catalytic properties of recombinant His6-GlpO from Mycoplasma pneumoniae (His6-MpGlpO) were investigated through transient and
Susan K Boehlein et al.
The Plant journal : for cell and molecular biology, 99(1), 23-40 (2019-02-13)
Cereal yields decrease when grain fill proceeds under conditions of prolonged, moderately elevated temperatures. Endosperm-endogenous processes alter both rate and duration of dry weight gain, but underlying mechanisms remain unclear. Heat effects could be mediated by either abnormal, premature cessation

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service