Skip to Content
Merck
All Photos(3)

Key Documents

G6264

Sigma-Aldrich

Guanosine

BioReagent, suitable for cell culture

Synonym(s):

9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
Beilstein:
625911
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

BioReagent

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

250 °C (dec.) (lit.)

solubility

0.05 M acetic acid: 1 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

[H]O[H].NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1

InChI key

NYHBQMYGNKIUIF-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine has been used:
  • as a ribonucleoside component of the basal medium to study its effects on Bacillus mojavensis′s anaerobic growth
  • as a component of the Dulbecco′s modified Eagle medium (DMEM) to culture mouse-derived embryonic stem cells
  • as a component of the nucleoside solution to culture chicken embryonic stem cells

Biochem/physiol Actions

Guanosine is a purine nucleoside that exhibits important functions in cell metabolism. It has also shown neuroprotective and neurotrophic effects in glial cells, hippocampal neurons, and pheochromocytoma cells by inducing proliferation and differentiation and also exerting anti-apoptotic effects. Guanosine acts as a therapeutic agent against several neuropathologies. It exerts its effects against sepsis-induced cognitive impairment, hepatic encephalopathy, lipopolysaccharide (LPS)-induced inflammation, ischemic injury, and azide-induced oxidative damage. Guanosine is suitable for use in cell culture applications as a precursor of guanosine monophosphate (GMP), guanosine diphosphate (GDP), guanosine triphosphate (GTP), and cyclic GMP (cGMP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffery T Davis et al.
Chemical Society reviews, 36(2), 296-313 (2007-02-01)
Nature's use of a simple genetic code to enable life's complex functions is an inspiration for supramolecular chemistry. DNA nucleobases carry the key information utilizing a variety of cooperative and non-covalent interactions such as hydrophobic, van der Waals, pi-pi stacking
Zoran D Matović et al.
Journal of inorganic biochemistry, 121, 134-144 (2013-02-05)
Novel square-planar palladium(II) complexes with O-N-N-O-type ligands H4mda (H4mda=malamido-N,N'-diacetic acid) and H4obp (H4obp=oxamido-N,N'-di-3-propionic acid) were prepared and characterized. The ligands coordinate to the palladium(II) ion via two pairs of deprotonated ligating atoms with square chelation. A four coordinate, square-planar geometry
Nadine Gehrke et al.
Immunity, 39(3), 482-495 (2013-09-03)
Immune sensing of DNA is critical for antiviral immunity but can also trigger autoimmune diseases such as lupus erythematosus (LE). Here we have provided evidence for the involvement of a damage-associated DNA modification in the detection of cytosolic DNA. The oxidized base
Aditya C Bandekar et al.
Current biology : CB, 30(20), 3961-3971 (2020-09-12)
Although the major events in prokaryotic cell cycle progression are likely to be coordinated with transcriptional and metabolic changes, these processes remain poorly characterized. Unlike many rapidly growing bacteria, DNA replication and cell division are temporally resolved in mycobacteria, making
Thomas Gonatopoulos-Pournatzis et al.
The Biochemical journal, 457(2), 231-242 (2013-12-21)
The 7mG (7-methylguanosine cap) formed on mRNA is fundamental to eukaryotic gene expression. Protein complexes recruited to 7mG mediate key processing events throughout the lifetime of the transcript. One of the most important mediators of 7mG functions is CBC (cap-binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service