Skip to Content
Merck
All Photos(1)

Key Documents

G4923

Sigma-Aldrich

(E)-Guggulsterone

≥95% (HPLC), powder

Synonym(s):

(17E)-Pregna-4,17(20)-diene-3,16-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: soluble 5 mg/mL

functional group

ketone

shipped in

ambient

storage temp.

2-8°C

SMILES string

C\C=C1\C(=O)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/b16-4-/t15-,17+,18+,20+,21-/m1/s1

InChI key

WDXRGPWQVHZTQJ-AUKWTSKRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Naturally occuring bioactive plant sterol isolated from the gum resin of guggul (Commiphora wightii). Antihyperlipidemic. Induces the expression of human CYP3A.. Guggulsterone potently reverses multi-drug resistance in a number of human cancer cell lines, extending the efficacy of chemotherapy.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang-Jip Nam et al.
Journal of natural products, 70(11), 1691-1695 (2007-11-09)
Three new scalarane-based sesterterpenes, 1- 3, were isolated from a marine sponge of the genus Spongia, and their chemical structures were elucidated by analysis of HRMS and 2-D NMR spectra. The isolated compounds 1 and 3 showed inhibition against the
Inas Almazari et al.
Topics in current chemistry, 329, 35-60 (2012-08-02)
Guggulsterone (GS) is a phytosterol derived from the gum resin of guggul plants that have been used traditionally to treat various disorders such as burns, wounds, gastric ulcer, cough, gum diseases, urinary complaints, diarrhea, stomach cramps, fascioliasis, and intestinal worms.
Dongfang Yang et al.
Journal of lipid research, 53(3), 529-539 (2012-01-17)
Z-Guggulsterone is a major ingredient in the Indian traditional hypolipidemic remedy guggul. A study in mice has established that its hypolipidemic effect involves the farnesoid X receptor (FXR), presumably by acting as an antagonist of this receptor. It is generally
Chunhua Zhu et al.
Lipids in health and disease, 10, 199-199 (2011-11-08)
Apolipoprotein M (apoM) may have potential antiatherosclerotic properties. It has been reported that apoM expression could be regulated by many intracellar and extracellar factors. In the present study we further investigated regulation of apoM expression in Caco-2 cells stimulated by
Jae-Jun Song et al.
International journal of pediatric otorhinolaryngology, 74(12), 1384-1387 (2010-10-26)
Guggulsterone is a bioactive constituent of resinous sap originating from the guggul tree, Commiphora mukul, which has been used over several thousands of years to treat various diseases, including atherosclerosis, rheumatism, and obesity. However, the effect of guggulsterone inflammatory reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service