Skip to Content
Merck
All Photos(1)

Key Documents

G4401

Sigma-Aldrich

Guanosine 5′-diphospho-β-L-fucose sodium salt

≥85%, powder

Synonym(s):

6-Deoxy-β-L-galactopyranosylguanosine 5′-diphosphate, GDP-Fuc, GDP-fucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23N5Na2O15P2
CAS Number:
Molecular Weight:
633.31
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Guanosine 5′-diphospho-β-L-fucose sodium salt, ≥85%

Quality Level

Assay

≥85%

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC1OC(OP(O)(=O)OP(O)(=O)OCC2OC(C(O)C2O)n3cnc4C(=O)N=C(N)Nc34)C(O)C(O)C1O

InChI

1S/C16H25N5O15P2/c1-4-7(22)9(24)11(26)15(33-4)35-38(30,31)36-37(28,29)32-2-5-8(23)10(25)14(34-5)21-3-18-6-12(21)19-16(17)20-13(6)27/h3-5,7-11,14-15,22-26H,2H2,1H3,(H,28,29)(H,30,31)(H3,17,19,20,27)

InChI key

LQEBEXMHBLQMDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanosine 5′-diphospho-β-L-fucose (GDP-fucose) is a sugar nucleotide present in bacteria and humans and is a fucosyltransferase substrate.

Application

Guanosine 5′-diphospho-β-L-fucose sodium salt may be used as a component of fucosyltransferase VII (FTVII) reaction buffer to aid exofucosylation in murine adipose tissue-derived mesenchymal stromal cells (AMSCs). It has been used for microbe agglutination assay and agglutination inhibition assay.
Guanosine 5′-diphospho-β-L-fucose sodium salt has been used for microbe agglutination assay and agglutination inhibition assay.

Substrates

Substrate for fucosyltransferase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Barbé et al.
Scientific reports, 8(1), 12961-12961 (2018-08-30)
Human strains of rotavirus A (RVAs) recognize fucosylated glycans belonging to histo-blood group antigens (HBGAs) through their spike protein VP8*. Lack of these ligands due to genetic polymorphisms is associated with resistance to gastroenteritis caused by P[8] genotype RVAs. With
Wei Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(38), 16096-16101 (2009-10-07)
Lewis X (Le(x))-containing glycans play important roles in numerous cellular processes. However, the absence of robust, facile, and cost-effective methods for the synthesis of Le(x) and its structurally related analogs has severely hampered the elucidation of the specific functions of
Daniel C Turnock et al.
The Journal of biological chemistry, 282(39), 28853-28863 (2007-07-21)
The protozoan parasite Trypanosoma brucei causes human African sleeping sickness in sub-Saharan Africa. The parasite makes several essential glycoproteins, which has led to the investigation of the sugar nucleotides and glycosyltransferases required to synthesize these structures. Fucose is a common
Diego Sánchez-Martínez et al.
Clinical and translational medicine, 11(2), e280-e280 (2021-02-27)
CD19-directed chimeric antigen receptors (CAR) T cells induce impressive rates of complete response in advanced B-cell malignancies, specially in B-cell acute lymphoblastic leukemia (B-ALL). However, CAR T-cell-treated patients eventually progress due to poor CAR T-cell persistence and/or disease relapse. The
David García-Bernal et al.
Frontiers in cell and developmental biology, 8, 584074-584074 (2020-12-17)
Mesenchymal stromal cells (MSCs) constitute the cell type more frequently used in many regenerative medicine approaches due to their exclusive immunomodulatory properties, and they have been reported to mediate profound immunomodulatory effects in vivo. Nevertheless, MSCs do not express essential

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service