Skip to Content
Merck
All Photos(1)

Key Documents

G2269

Sigma-Aldrich

Gastric Inhibitory Polypeptide human

≥95% (HPLC), suitable for ligand binding assays

Synonym(s):

Glucose-dependent Insulinotropic Peptide, GIP, Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C226H338N60O66S
CAS Number:
Molecular Weight:
4983.53
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Gastric Inhibitory Polypeptide human, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

[H]N[C@H](C(N[C@H](C(N[C@@H](CCC(O)=O)C(NCC(N[C@]([C@H](O)C)([H])C(N[C@H](C(N[C@]([C@@H](C)CC)([H])C(N[C@@H](CO)C(N[C@H](C(N[C@H](C(N[C@@H](CO)C(N[C@]([C@@H](C)CC)([H])C(N[C@H](C(N[C@H](C(N[C@H](C(N[C@@H](CCCCN)C(N[C@]([C@@H](C)CC)([H])C(N[C@H](C(N[C@@H](

InChI key

MGXWVYUBJRZYPE-YUGYIWNOSA-N

Gene Information

human ... GIP(2695)

Amino Acid Sequence

Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln

Biochem/physiol Actions

Gastric Inhibitory Polypeptide (GIP), a secretin family incretin hormone, is produced by K-cells resident in the gastrointestinal tract and within the CNS. GIP is use to study its diverse receptor mediated functions that range from weight loss to neuronal survival.

Other Notes

InChI: 1S/C226H338N60O66S/c1-21-113(11)181(285-218(343)165(107-288)278-203(328)150(88-125-60-64-131(292)65-61-125)264-212(337)163(99-179(310)311)274-217(342)164(106-287)279-222(347)183(115(13)23-3)283-215(340)152(87-123-47-29-26-30-48-123)275-224(349)185(120(18)289)280-174(301)105-245-192(317)142(70-75-175(302)303)252-187(312)117(15)248-190(315)134(232)85-124-58-62-130(291)63-59-124)220(345)250-119(17)189(314)254-146(76-82-353-20)199(324)272-160(96-176(304)305)210(335)258-141(57-39-44-81-231)200(325)282-182(114(12)22-2)221(346)276-156(92-129-103-241-109-247-129)206(331)260-144(67-72-167(234)294)197(322)259-145(68-73-168(235)295)198(323)271-161(97-177(306)307)211(336)265-151(86-122-45-27-25-28-46-122)214(339)281-180(112(9)10)219(344)277-158(94-171(238)298)209(334)266-154(90-127-101-243-136-52-34-32-50-133(127)136)205(330)263-149(84-111(7)8)202(327)262-148(83-110(5)6)201(326)249-118(16)188(313)253-143(66-71-166(233)293)196(321)255-137(53-35-40-77-227)191(316)244-104-173(300)251-138(54-36-41-78-228)193(318)256-139(55-37-42-79-229)195(320)269-157(93-170(237)297)208(333)273-162(98-178(308)309)213(338)267-153(89-126-100-242-135-51-33-31-49-132(126)135)204(329)257-140(56-38-43-80-230)194(319)268-155(91-128-102-240-108-246-128)207(332)270-159(95-172(239)299)216(341)284-184(116(14)24-4)223(348)286-186(121(19)290)225(350)261-147(226(351)352)69-74-169(236)296/h25-34,45-52,58-65,100-103,108-121,134,137-165,180-186,242-243,287-292H,21-24,35-44,53-57,66-99,104-107,227-232H2,1-20H3,(H2,233,293)(H2,234,294)(H2,235,295)(H2,236,296)(H2,237,297)(H2,238,298)(H2,239,299)(H,240,246)(H,241,247)(H,244,316)(H,245,317)(H,248,315)(H,249,326)(H,250,345)(H,251,300)(H,252,312)(H,253,313)(H,254,314)(H,255,321)(H,256,318)(H,257,329)(H,258,335)(H,259,322)(H,260,331)(H,261,350)(H,262,327)(H,263,330)(H,264,337)(H,265,336)(H,266,334)(H,267,338)(H,268,319)(H,269,320)(H,270,332)(H,271,323)(H,272,324)(H,273,333)(H,274,342)(H,275,349)(H,276,346)(H,277,344)(H,278,328)(H,279,347)(H,280,301)(H,281,339)(H,282,325)(H,283,340)(H,284,341)(H,285,343)(H,286,348)(H,302,303)(H,304,305)(H,306,307)(H,308,309)(H,310,311)(H,351,352)/t113-,114-,115-,116-,117-,118-,119-,120+,121+,134-,137-,138-,139-,140-,141-,142-,143-,144-,145-,146-,147-,148-,149-,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,180-,181-,182-,183-,184-,185-,186-/m0/s1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a 42 residue peptide corresponding to the entire amino acid sequence of human GIP.
H Yajima et al.
Chemical & pharmaceutical bulletin, 33(8), 3578-3581 (1985-08-01)
Diabetes remission after bariatric surgery: is it just the incretins?
Laferrere B.
International Journal of Obesity, 35, S22-S25 (2011)
Sabrina Paratore et al.
Central nervous system agents in medicinal chemistry, 11(3), 210-222 (2011-09-17)
The development of neuronal apoptosis depends on an intrinsic transcriptional program. By DNA microarray technology, we have previously implicated a number of genes in different paradigms of neuronal apoptosis. In the present study, we investigated the spatiotemporal pattern of expression
Priya Sumithran et al.
The New England journal of medicine, 365(17), 1597-1604 (2011-10-28)
After weight loss, changes in the circulating levels of several peripheral hormones involved in the homeostatic regulation of body weight occur. Whether these changes are transient or persist over time may be important for an understanding of the reasons behind
A J Moody et al.
FEBS letters, 172(2), 142-148 (1984-07-09)
Human GIP 1-42 and fragments of human GIP corresponding to GIP 10-42, GIP 11-42, and GIP 17-42 were isolated from acid-ethanol extracts of human small intestines with the aid of an anti-GIP serum specific for the extreme C-terminal portion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service