Skip to Content
Merck
All Photos(1)

Key Documents

G1549

Sigma-Aldrich

PNGase F from Elizabethkingia meningoseptica

ready-to-use solution, recombinant, expressed in E. coli

Synonym(s):

PNGase F from Elizabethkingia meningoseptica, N-Glycosidase F, PNGase F from Chryseobacterium meningosepticum, PNGase F from Flavobacterium meningosepticum, Peptide N-glycosidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

grade

Proteomics Grade

form

ready-to-use solution

specific activity

≥1000 U/mg

shelf life

≥1 yr at -20 °C

mol wt

~36 kDa

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Recombinant PNGase F has been purified by affinity chromatography and dialyzed into a 50% glycerol solution with 10 mM potassium phoosphate pH 7.5 to produce a stable product. The product contains low levels of buffer salts. This highly purified material can be used for preparative deglycosylation or for analytical applications in gel, in solution, or on blot membranes. The enzyme can be removed from preparative operations by utilizing its C-terminal 6x histidine fusion tag. PNGase F from Elizabethkingia meningoseptica has been used in deglycosylation assay in human plasma samples and in deglycosylation of chondroitin sulfate proteoglycan.
Used to deglycosylate protein.

Biochem/physiol Actions

PNGase F from Elizabethkingia meningoseptica has glycan-binding catalytic domain and a bowl-like domain at the N-terminus. It cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain. It is cost-effectively produced on a large scale in prokaryotic hosts and requires divalent zinc ions for its enzymatic activity.
Cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain.

Unit Definition

One unit will catalyze the release of N-linked oligosaccharides from 1 nanomole of denatured ribonuclease B in one minute at 37°C at pH 7.5 monitored by SDS-PAGE. One Sigma unit of PNGase F activity is equal to 1 IUB milliunit.

Physical form

Supplied as 300 Units/mL enzyme in 50% (v/v) glycerol and 50% (v/v) 20 mM Potassium Phosphate, pH 7.5.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-glycosylation of apolipoprotein A1 in cardiovascular diseases
Majek P, et al.
Translational Research, 165(2), 360-362 (2015)
Characterization and analysis of extracellular matrix in malignant brain tumors and their cellular derivatives
Extracellular Matrix, 113-138 (2015)
Identification and characterization of a novel prokaryotic peptide: N-glycosidase from Elizabethkingia meningoseptica
Sun G, et al.
The Journal of Biological Chemistry, jbc-M114 (2015)
Discovery and characterization of a novel extremely acidic bacterial N-glycanase with combined advantages of PNGase F and A
Wang T, et al.
Bioscience Reports, 34(6), e00149-e00149 (2014)
T H Plummer et al.
The Journal of biological chemistry, 259(17), 10700-10704 (1984-09-10)
Endo-beta-N-acetylglucosaminidase F preparations from Flavobacterium meningosepticum have been found to contain peptide:N-glycosidase activity. Only the second activity, designated as peptide:N-glycosidase F, readily cleaves the beta-aspartylglycosylamine linkage of a fetuin triantennary complex glycopeptide, as shown by the isolation of the corresponding

Articles

Antibody fragmentation with our pepsin digestion protocol for IgG antibody fragmentation and preparation of F(ab’).

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service