Skip to Content
Merck
All Photos(3)

Key Documents

G1152

Sigma-Aldrich

Glass beads, acid-washed

710-1,180 μm (16-25 U.S. sieve)

Synonym(s):

Glass Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.22

particle size

710-1,180 μm (16-25 U.S. sieve)

Application

Acid-washed glass beads have been used:
  • in the extraction of proteins from antigen-expressing transgenic lines of Nicotiana tabacum L.
  • to extract DNA of Candida albicans from human and murine blood samples
  • to measure lipid yield from microalga C.homosphaera by adopting different methods of cell disruption methods

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarina Damjanovic et al.
Microbial ecology, 79(3), 706-719 (2019-08-23)
Coral-associated bacteria are critical for the well-being of their host and may play essential roles during ontogeny, as suggested by the vertical transmission of some bacteria in brooding corals. Bacterial acquisition patterns in broadcast spawners remain uncertain, as 16S rRNA
Continued expression of plant-made vaccines following long-term cryopreservation of antigen-expressing tobacco cell cultures
Van E J and Keen P.
In Vitro Cellular & Developmental Biology. Plant, 45(6), 750-757 (2009)
Novel strategy for microalgae cell disruption and wet lipid extraction by employing electro-Fenton process with sacrificial steel anode
Wanniarachchige P, et al.
Bioresource Technology (2022)
Evaluation of a digital microfluidic real-time PCR platform to detect DNA of Candida albicans in blood
W. A. Schell
European Journal of Clinical Microbiology & Infectious Diseases : Official Publication of the European Society of Clinical Microbiology (2012)
Sarah L Sokol et al.
eLife, 7 (2018-05-23)
Most eukaryotic parasites are obligately heteroxenous, requiring sequential infection of different host species in order to survive. Toxoplasma gondii is a rare exception to this rule, having a uniquely facultative heteroxenous life cycle. To understand the origins of this phenomenon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service