Skip to Content
Merck
All Photos(1)

Documents

G0411

Sigma-Aldrich

Prionex® Highly purified Type A

aqueous solution

Synonym(s):

Gelatin, Prionex® Highly purified Type A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

Porcine bone

Quality Level

sterility

aseptically processed

form

aqueous solution

technique(s)

ELISA: suitable
microbiological culture: suitable
western blot: suitable

General description

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, bones, etc. in water. Type-A gelatin is derived from acid-cured (acid-hydrolyzed) tissue. Type-B gelatin is derived from lime-cured (base/alkaline-hydrolyzed) tissue. Gelatin is a biocompatible polymer.

Application

Prionex® Highly purified Type A has been used:
  • for coating cell culture to improve attachment of cells, and was added to polymerase chain reaction (PCR) to help stabilize Taq DNA
  • as a blocking reagent in Western blotting, enzyme linked immunosorbent assay (ELISA), and immunohistochemistry
  • as a component of media for species differentiation in bacteriology
  • as a delivery vehicle for the release of bioactive molecules and in the generation of scaffolds for engineering applications
  • in the murine lethality assay which is used to examine the ability of SMAbs to neutralize the effects of BoNT/A1 in vivo

Legal Information

Prionex is a registered trademark of Pentapharm AG, Basel

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino Acid Composition of Gelatin from Ephinephelus sp.
Siregar GRM and Suprayitno E
IOSR Journal of Agriculture and veterinary Science, 12(4), 51-54 (2019)
Gels for Photographic Emulsions
Encyclopedia of Materials: Science and Technology, 3495-3497 (2001)
PCR Kit
PCR Primer: A Laboratory Manual (1995)
Photographic Processes and Materials
Encyclopedia of Physical Science and Technology, 91-132 (2001)
Kevin Retailleau et al.
Toxins, 14(3) (2022-03-25)
For the past two decades, botulinum neurotoxin A (BoNT/A) has been described as a strong candidate in the treatment of pain. With the production of modified toxins and the potential new applications at the visceral level, there is a real

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service