Skip to Content
Merck
All Photos(1)

Key Documents

F8307

Sigma-Aldrich

FR-900098 monosodium salt

≥97% (NMR)

Synonym(s):

(3-(Acetylhydroxyamino)propyl)-phosphonic acid, BRN 2096083, P-[3-(Acetylhydroxyamino)propyl]-phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NNaO5P
CAS Number:
Molecular Weight:
219.11
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (NMR)

form

powder

storage condition

desiccated
protect from light

color

white to light brown

solubility

H2O: ≥20 mg/mL

storage temp.

−20°C

SMILES string

[Na+].CC(=O)N(O)CCCP(O)([O-])=O

InChI

1S/C5H12NO5P.Na/c1-5(7)6(8)3-2-4-12(9,10)11;/h8H,2-4H2,1H3,(H2,9,10,11);/q;+1/p-1

InChI key

LCFXFDGYDKNWMD-UHFFFAOYSA-M

Biochem/physiol Actions

1-deoxy-D-xylulose 5-phosphate reductoisomerase (DXR) is an enzyme involved in the first step in the nonmevalonate pathway for isoprenoid biosynthesis in Gram-negative, Gram-positive bacteria, plants, and the parasite causing the most virulent form of malaria, Plasmodium falciparum (Mammals produce isoprenoids via the mevalonate pathway). Inhibiton of the enzyme provides alternative to traditional antimalaria therapy. FR-900098 is twice more effective than fosmidomycin against various strains of P. falciparum in vitro and the closely related P. vinckei in mice.
1-deoxy-D-xylulose 5-phosphate reductoisomerase (DXR) is an enzyme involved in the first step in the nonmevalonate pathway for isoprenoid biosynthesis in Gram-negative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service