Skip to Content
Merck
All Photos(1)

Key Documents

F7759

Sigma-Aldrich

Anti-Horse IgG (whole molecule)−FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Horse IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: suitable

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

An immunoglobulin has two heavy chains and two light chains connected by disulfide bond. It is a glycoprotein. IgG is a major class of immunoglobulin. Horse IgG has different subclasses, IgG2a, IgG2b, IgG2c, IgG1 and IgG(T). Compared to other IgG molecules, IgG(T) has more carbohydrate content. In healthy individuals, total IgG is dispersed equally between circulating plasma and interstitial fluids.
Horse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in horse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Specificity

Anti-horse IgG (whole molecule)-FITC antibody is specific for horse IgG subclasses.

Immunogen

Purified horse IgG

Application

Anti-Horse IgG (whole molecule) FITC antibody produced in rabbit has been used in indirect fluorescent antibody test. It has also been used as a secondary antibody to confirm equine arteritis virus (EAV).
Rabbit polyclonal anti-Horse IgG (whole molecule)−FITC antibody may be used to detect and quantitate the level of IgG in horse serum and biological fluids by fluorescent techniques. It may also be used as a secondary antibody in assays that use horse IgG as the primary antibody.

Biochem/physiol Actions

IgG antibody subtype is the most abundant serum immunoglobulin of the immune system. It is secreted by B- cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to the fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. It mainly helps in immune defense.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A 34-year retrospective study of equine viral abortion in Poland
Bazanow BA, et al.
Polish Journal of Veterinary Sciences, 17(4), 607-612 (2014)
Alessia Libera Gazzonis et al.
Parasites & vectors, 13(1), 530-530 (2020-10-24)
Leishmania infantum is a vector-borne pathogen endemic in countries in the Mediterranean basin, including Italy. Dogs act as the primary reservoir for this parasite, but other animal species may also be infected. Low-to-moderate seroprevalence levels of infection have been reported
Alexandre Dias Munhoz et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 28(2), 283-290 (2019-06-20)
The aim of this study was to determine the frequency and factors associated with Toxoplasma gondii in naturally infected equids in northeastern Brazil. Serum samples from 569 equids (528 horses, 8 mules and 33 donkeys) were subjected to the indirect
Samantha Gusmão Pellizzoni et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 30(2), e027720-e027720 (2021-06-24)
The aims of this study were to determine the seroprevalence of Sarcocystis neurona antibodies in equines in the Ilhéus-Itabuna microregion (BA), and identify possible factors associated with infection. The presence of sporocysts/oocysts of Sarcocystis spp. was also verified in Didelphis
B A Bażanów et al.
Polish journal of veterinary sciences, 17(4), 607-612 (2015-02-03)
The purpose of the present review was a comparison of the abortions caused by EAV and EHV-1 viruses over the 34 years. A total of 452 tissues samples from aborted fetuses (347) or foals (105) stillborn or newborn that died

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service