Skip to Content
Merck
All Photos(1)

Key Documents

F5512

Sigma-Aldrich

Anti-Human IgG (Fab specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (Fab specific)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunofluorescence: 1:16-1:32 using Hep2 cells

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein and mainly helps in immune defense. IgG is usually found as a monomer. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4. About 70 percent of the total immunoglobulin consists of IgG. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III.
The product binds all human Igs and provides reduced background with mouse or rat samples.

Application

Anti-Human IgG (Fab specific)-FITC antibody has been used:
  • in fluorescent staining
  • in dual color fluorescent labeling
  • in fluorescence-activated cell sorting (FACS) analysis

Anti-Human IgG (Fab specific)-FITC antibody produced in goat may be used at a working antibody dilution of 1:16 - 1:32 for anti-nuclear antibody (A.N.A.) assay on acetone-fixed human Hep2 cells. This antibody was used in FACS of HNE2 and KNE2/LMP1 cells. It was also used to perform TARDIS assay for detection of topoisomerases I and II that are covalently bound to DNA in K562 leukemia cells.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of FITC to Anti- Human IgG (Fab specific) antibody allows for the visualization of protein by fluorescent microscopy.

Other Notes

No cross-reaction with mouse and rat IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular imaging application of radioiodinated anti-EGFR human Fab to EGFR-overexpressing tumor xenografts
Xu N, et al.
Anticancer research, 29(10), 4005-4011 (2009)
A human Fab exclusively binding to the extracellular domain of LMP2A
Cao Q, et al.
Biochemical and biophysical research communications, 482(2), 226-231 (2017)
Renjie Chen et al.
Molecular cancer therapeutics, 11(3), 594-603 (2011-12-16)
Nasopharyngeal carcinoma (NPC) is a major cause of cancer-related death in Southeast Asia and China. Metastasis and relapse are the primary cause of morbidity and mortality in NPC. Recent evidence suggests that the Epstein-Barr virus latent membrane protein 1 (LMP1)
Hiroyuki Takamatsu et al.
Journal of immunology (Baltimore, Md. : 1950), 182(1), 703-710 (2008-12-26)
Moesin is an intracellular protein that links the cell membrane and cytoskeleton, while also mediating the formation of microtubules and cell adhesion sites as well as ruffling of the cell membrane. To determine the roles of anti-moesin Abs derived from
Xin Wang et al.
Cancer biology & therapy, 6(6), 980-987 (2007-05-31)
The aims of this study were to generate a human Fab fragment against EGFR; conjugate it to paclitaxel (Taxol) as an immuno-chemotherapy agent; and investigate its in vitro anti-tumor efficacy on A431 epidermoid carcinoma cells. A431 cells (EGFR-positive), NIH 3T3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service