Skip to Content
Merck
All Photos(1)

Key Documents

F4505

Sigma-Aldrich

m-Fluoro-DL-tyrosine

≥98%, suitable for ligand binding assays

Synonym(s):

3-fluoro-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3-4-(OH)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
199.18
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

m-Fluoro-DL-tyrosine,

Assay

≥98%

form

powder

technique(s)

ligand binding assay: suitable

color

white

mp

280 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

NC(Cc1ccc(O)c(F)c1)C(O)=O

InChI

1S/C9H10FNO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)

InChI key

VIIAUOZUUGXERI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

M-Fluorotyrosine (m-Fluoro-DL-tyrosine) may be substituted for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, eg. aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Jordan et al.
Brain research, 750(1-2), 264-276 (1997-03-07)
The tracer 6-[18F]fluoro-L-m-tyrosine (FMT) was studied with regard to its biochemistry and kinetics, as well as its utility in evaluating brain dopaminergic function in primates before and after 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) treatment using positron emission tomography (PET). Plasma analysis of FMT
Steven Y Reece et al.
Journal of the American Chemical Society, 128(42), 13654-13655 (2006-10-19)
The complex Re(bpy)(CO)3CN is an excited state oxidant of tyrosine upon deprotonation of the tyrosyl phenol. A series of Re(bpy-FnY)(CO)3CN complexes ([Re]-FnY: [Re]-Y, [Re]-3-FY, [Re]-3,5-F2Y, [Re]-2,3-F2Y, [Re]-2,3,5-F3Y, [Re]-2,3,6-F3Y, and [Re]-F4Y) have been prepared so as to vary the FnY*/FnY- reduction
Julianne L Kitevski-LeBlanc et al.
Journal of biomolecular NMR, 45(3), 255-264 (2009-08-06)
Fluorine NMR is a useful tool to probe protein folding, conformation and local topology owing to the sensitivity of the chemical shift to the local electrostatic environment. As an example we make use of (19)F NMR and 3-fluorotyrosine to evaluate
Crystallographic evidence for isomeric chromophores in 3-fluorotyrosyl-green fluorescent protein.
Jae Hyun Bae et al.
Chembiochem : a European journal of chemical biology, 5(5), 720-722 (2004-05-04)
Marie-Claude Asselin et al.
Physics in medicine and biology, 47(11), 1961-1977 (2002-07-11)
Physiological quantification of dynamic PET data requires the determination of an input function, preferably from plasma. A compartmental model relating a parent radiotracer, its radiolabelled metabolites and their exchange between plasma and erythrocytes is presented. This model allows for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service